Skip to Content
Merck
All Photos(1)

Key Documents

O9387

Sigma-Aldrich

Oxatomide

≥99%

Synonym(s):

1-[3-[4-(Diphenylmethyl)-1-piperazinyl]propyl]-1,3-dihydro-2H-benzimidazol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30N4O
CAS Number:
Molecular Weight:
426.55
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥99%

form

powder

color

white

solubility

DMSO: soluble
ethanol: soluble

originator

Johnson & Johnson

SMILES string

O=C1Nc2ccccc2N1CCCN3CCN(CC3)C(c4ccccc4)c5ccccc5

InChI

1S/C27H30N4O/c32-27-28-24-14-7-8-15-25(24)31(27)17-9-16-29-18-20-30(21-19-29)26(22-10-3-1-4-11-22)23-12-5-2-6-13-23/h1-8,10-15,26H,9,16-21H2,(H,28,32)

InChI key

BAINIUMDFURPJM-UHFFFAOYSA-N

Gene Information

human ... DRD3(1814)

General description

Oxatomide, a histamine H1-receptor antagonist, is bound at high levels to plasma proteins in human blood.

Biochem/physiol Actions

Oxatomide, found in several antihistamines, can suppress mast cell degranulation. It can be used as an alternative to individuals with allergic rhinitis who do not or poorly respond to more established antihistamines. Oxatomide exhibits both antihistamine and anti-inflammatory properties. It exhibits therapeutic effects against asthma and vulvar lichen sclerosus.
Oxatomide is an anti-allergy compound. It suppresses platelet activating factor (PAF)-induced bronchoconstriction and inhibits leukotriene production.

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariko Domae et al.
The Journal of allergy and clinical immunology, 111(3), 567-572 (2003-03-19)
Eosinophils accumulated in sites of allergic inflammation are thought to play a crucial role in the pathogenesis of allergic disorders including asthma, allergic rhinitis, and atopic dermatitis, and tissue eosinophilia is attributable to increased eosinophil survival or decreased eosinophil apoptosis.
H Kimata
European journal of clinical investigation, 33(6), 513-517 (2003-06-11)
Playing video games causes physical and psychological stress, including increased heart rate and blood pressure and aggression-related feelings. Use of mobile phones is very popular in Japan, and frequent ringing is a common and intrusive part of Japanese life. Atopic
Hiroyuki Mizuguchi et al.
Pharmaceutics, 12(6) (2020-06-12)
H1-antihistamines antagonize histamine and prevent it from binding to the histamine H1 receptor (H1R). Some of them also act as inverse agonists, which are more potent than pure antagonists because they suppress the constitutive H1R activity. Bilastine is a non-sedative
D M Richards et al.
Drugs, 27(3), 210-231 (1984-03-01)
Oxatomide is an orally active H1-histamine receptor antagonist which, as appears to occur with some other antihistamines, also inhibits mast cell degranulation. Oxatomide has demonstrated response rates similar to those with other more established members of its drug class in
C Dani et al.
Drugs under experimental and clinical research, 28(5), 207-210 (2003-03-15)
The pharmacokinetics and tolerability of oxatomide oral suspension were investigated in preterm infants to evaluate the feasibility of planning a further study to assess its antiinflammatory effects and its effectiveness in preventing chronic lung disease (CLD). Following the administration of

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service