Skip to Content
Merck
All Photos(1)

Key Documents

P0044

Sigma-Aldrich

Phosphatase Inhibitor Cocktail 3

DMSO solution, for the inhibition of protein phosphatase 2A, alkaline phosphatases, protein phosphatases 1 and 2A, DMSO solution

Synonym(s):

Phosphatase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77
form:
DMSO solution
solubility:
water: soluble
storage temp.:
2-8°C

Product Name

Phosphatase Inhibitor Cocktail 3, DMSO solution

form

DMSO solution

Quality Level

solubility

water: soluble

shipped in

wet ice

storage temp.

2-8°C

Application

The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts. This product has been tested on extract from animal tissues including human placenta and bovine liver, and on extracts of A431, CHO, and U937 cells. It has been optimized and tested for L-isozymes of alkaline phosphatase as measured with pNPP and for serine-threonine protein phosphatases.

Biochem/physiol Actions

This mixture contains individual components with specific inhibitory properties. Cantharidin inhibits protein phosphatase 2A. (-)-p-Bromolevamisole oxalate inhibits L-isoforms of alkaline phosphatases. Calyculin A inhibits protein phosphatases 1 and 2A.

Caution

As supplied, this product is stable for four years at the recommended 2-8°C storage temperature.

Preparation Note

This cocktail is supplied as a clear solution in dimethyl sulfoxide. It is recommended to dilute the cocktail at 1 mL of cocktail per 500 mg of protein extracted from tissue or cells. Often a final concentration of 1% (v/v) is used, which will inhibit phosphatase activities found in the 27,000 x g supernatant from human placenta and bovine liver, and in the 100,000 x g supernatant from A431, CHO, and U937 extracts.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinglin L Xie et al.
Nature communications, 8(1), 499-499 (2017-09-13)
The capacity to coordinate environmental sensing with initiation of cellular responses underpins microbial survival and is crucial for virulence and stress responses in microbial pathogens. Here we define circuitry that enables the fungal pathogen Candida albicans to couple cell cycle
Tetsuro Ikegami et al.
PLoS pathogens, 5(2), e1000287-e1000287 (2009-02-07)
Rift Valley fever virus (RVFV) (genus Phlebovirus, family Bunyaviridae) is a negative-stranded RNA virus with a tripartite genome. RVFV is transmitted by mosquitoes and causes fever and severe hemorrhagic illness among humans, and fever and high rates of abortions in
Sarahmi Ishida et al.
The Plant cell, 20(12), 3273-3288 (2008-12-25)
The homeostasis of gibberellins (GAs) is maintained by negative feedback in plants. REPRESSION OF SHOOT GROWTH (RSG) is a tobacco (Nicotiana tabacum) transcriptional activator that has been suggested to play a role in GA feedback by the regulation of GA
Shumei Ma et al.
PloS one, 12(8), e0182921-e0182921 (2017-08-23)
Ferroptosis is a cell death pathway characterized by iron-dependent accumulation of reactive oxygen species (ROS) within the cell. The combination of siramesine, a lysosome disruptor, and lapatinib, a dual tyrosine kinase inhibitor, has been shown to synergistically induce cell death
Cindy Puente et al.
The Journal of biological chemistry, 291(11), 6026-6035 (2016-01-24)
Autophagy is a conserved catabolic process that utilizes a defined series of membrane trafficking events to generate a de novo double-membrane vesicle termed the autophagosome, which matures by fusing to the lysosome. Subsequently, the lysosome facilitates the degradation and recycling

Articles

The use of phosphatase inhibitors is critical for all types of phosphorylation studies. Compare phosphatase inhibitor cocktails to select the best one for your research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service