Skip to Content
Merck
All Photos(2)

Documents

N3641

Sigma-Aldrich

4-Nitrophenyl α-L-arabinofuranoside

chromogenic, ≥98% (TLC), powder

Synonym(s):

4-nitrophenyl-ARA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO7
CAS Number:
Molecular Weight:
271.22
Beilstein:
1688357
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Nitrophenyl α-L-arabinofuranoside, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

solubility

methanol: 50 mg/mL

storage temp.

−20°C

SMILES string

OC[C@@H]1O[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@H]1O

InChI

1S/C11H13NO7/c13-5-8-9(14)10(15)11(19-8)18-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2/t8-,9-,10+,11+/m0/s1

InChI key

DUYYBTBDYZXISX-UKKRHICBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl α-L-arabinofuranoside has been used:
  • as a substrate for α-arabinofuranosidase in an enzymatic assay to determine glycosidase activities in Oenococcus oeni strains
  • to determine the enzymatic activity of α-arabinofuranosidase
  • as a p-nitrophenol linked substrate to determine its activity in Aspergillus niger NW249 culture filtrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liudmila V Kozlova et al.
Planta, 241(5), 1159-1172 (2015-01-23)
Specific α- l -arabinofuranosidases are involved in the realisation of elongation growth process in cells with type II cell walls. Elongation growth in a plant cell is largely based on modification of the cell wall. In type II cell walls
José Manuel Inácio et al.
Journal of bacteriology, 190(12), 4272-4280 (2008-04-15)
The extracellular depolymerization of arabinopolysaccharides by microorganisms is accomplished by arabinanases, xylanases, and galactanases. Here, we characterize a novel endo-alpha-1,5-l-arabinanase (EC 3.2.1.99) from Bacillus subtilis, encoded by the yxiA gene (herein renamed abn2) that contributes to arabinan degradation. Functional studies
Preparation of arabinoxylobiose from rye xylan using family 10 Aspergillus aculeatus endo-1, 4-beta-D-xylanase
Rantanen H, et al.
Carbohydrate Polymers null
M J Renner et al.
Applied and environmental microbiology, 64(1), 43-52 (1998-01-22)
An alpha-L-arabinofuranosidase (alpha-L-arabinofuranoside arabinofuranohydrolase [EC 3.2.1.55]; referred to below as ArfI) from Cytophaga xylanolytica XM3 was purified 85-fold by anion-exchange and hydrophobic interaction column chromatography. The native enzyme had a pI of 6.1 and an apparent molecular mass of 160
Andrew Diener
Molecular plant-microbe interactions : MPMI, 25(12), 1531-1541 (2012-08-17)
Host-specific forms of Fusarium oxysporum infect the roots of numerous plant species. I present a novel application of familiar methodology to visualize and quantify F. oxysporum in roots. Infection in the roots of Arabidopsis thaliana, tomato, and cotton was detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service