N2288
Nefiracetam
solid
Synonym(s):
(2-(2-Oxopyrrolidin-1-yl)-N-(2,6-dimethylphenyl)-acetamide), DM-9384
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H18N2O2
CAS Number:
Molecular Weight:
246.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
form
solid
color
white to off-white
solubility
DMSO: >5 mg/mL
SMILES string
Cc1cccc(C)c1NC(=O)CN2CCCC2=O
InChI
1S/C14H18N2O2/c1-10-5-3-6-11(2)14(10)15-12(17)9-16-8-4-7-13(16)18/h3,5-6H,4,7-9H2,1-2H3,(H,15,17)
InChI key
NGHTXZCKLWZPGK-UHFFFAOYSA-N
Biochem/physiol Actions
Nefiracetam (NEF) is a pyrrolidonetype nootropic agent with various pharmacologic as well as cognition enhancing effects. In amygdala-kindled seizures, nefiracetam inhibits both electroencephalographic and behavioral seizures. NEF has a distinct anticonvulsant spectrum.
Novel pyrrolidonetype nootropic (condition enhancing) agent with anticonvulsant spectrum.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Pharmacological study on Alzheimer's drugs targeting calcium/calmodulin-dependent protein kinase II.
Shigeki Moriguchi
Journal of pharmacological sciences, 117(1), 6-11 (2011-08-09)
In the brain of Alzheimer's disease patients, down-regulation of both cholinergic and glutamatergic systems have been found and is thought to play an important role in impairment of cognition, learning, and memory. Nefiracetam is a pyrrolidine-related nootropic drug exhibiting various
Shigeki Moriguchi et al.
Journal of neurochemistry, 106(3), 1092-1103 (2008-05-01)
Nefiracetam is a pyrrolidine-related nootropic drug exhibiting various pharmacological actions such as cognitive-enhancing effect. We previously showed that nefiracetam potentiates NMDA-induced currents in cultured rat cortical neurons. To address questions whether nefiracetam affects NMDA receptor-dependent synaptic plasticity in the hippocampus
Yoshimi Tsuchiya et al.
Toxicologic pathology, 33(5), 561-569 (2005-08-18)
To ascertain the early pathophysiological features in canine renal papillary necrosis (RPN) caused by the neurotransmission enhancer nefiracetam, male beagle dogs were orally administered nefiracetam at 300 mg/kg/day for 4 to 7 weeks in comparison with ibuprofen, a non-steroidal anti-inflammatory
Ricardo E Jorge et al.
Canadian journal of psychiatry. Revue canadienne de psychiatrie, 55(6), 350-354 (2010-06-15)
We will review the available evidence on the frequency, clinical correlates, mechanism, and treatment of apathy following stroke. We have explored relevant databases (that is, PubMed, MEDLINE, and PsycINFO) using the following key words and their combinations: apathy, motivation, abulia
Yijin Yan et al.
Behavioural brain research, 173(1), 39-46 (2006-07-22)
Animal models of drug discrimination have been used to examine the subjective effects of addictive substances. The cAMP system is a crucial downstream signaling pathway implicated in the long-lasting neuroadaptations induced by addictive drugs. We examined effects of rolipram, nefiracetam
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service