Skip to Content
Merck

GRSA

Glutathione Reductase Assay Kit

Sufficient for 100 colorimetric tests

Synonym(s):

Glutathione Reductase Activity Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.28
UNSPSC Code:
12161503

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

usage

sufficient for 100 tests

detection method

colorimetric

storage temp.

2-8°C

Gene Information

human ... GSR(2936)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CGP1CS0260CS0170
Gene Information

human ... GSR(2936)

Gene Information

-

Gene Information

human ... GSR(2936)

Gene Information

human ... TXNRD1(7296), TXNRD2(10587), TXNRD3(114112)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

detection method

colorimetric

detection method

colorimetric

detection method

-

detection method

-

usage

sufficient for 100 tests

usage

sufficient for 100 tests

usage

sufficient for 700 assays

usage

 kit sufficient for 100 assays (1 mL)

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

General description

Glutathione reductase (GR) is a ubiquitous enzyme that catalyzes the reduction of oxidized glutathione (GSSG) to glutathione (GSH). Glutathione reductase is essential for the glutathione redox cycle that maintains adequate levels of reduced cellular GSH, which serves as an antioxidant reacting with free radicals and organic peroxides. Glutathione is also a substrate for the glutathione peroxidases and glutathione S-transferases in the detoxification of organic peroxides and the metabolism of xenobiotics. 

Application

Suitable for the measurement of glutathione reductase activity in biological samples
Glutathione Reductase Assay Kit has been used to measure the activity of glutathione reductase as a part of oxidative stress assessment and also to study the effects of antifouling biocides on it.

Biochem/physiol Actions

Glutathione reductase (EC 1.6.4.2) (GR) is a ubiquitous enzyme that catalyzes the reduction of oxidized glutathione (GSSG) to glutathione (GSH). Glutathione reductase is essential for the glutathione redox cycle that maintains adequate levels of reduced cellular GSH, which serves as an antioxidant reacting with free radicals and organic peroxides. Glutathione is also a substrate for the glutathione peroxidases and glutathione S-transferases in the detoxification of organic peroxides and the metabolism of xenobiotics. This kit contains reagents for the spectrophotometric determination of glutathione reductase activity either by following the decrease in absorbance caused by the oxidation of NADPH at 340 nm (UV assay) or the increase in absorption caused by the reduction of dithiobis(2-nitrobenzoic acid) (DTNB) at 412 nm (colorimetric assay). This kit provides reagents for a spectrophotometric assay for measuring the activity of glutathione reductase either by following the decrease in A340 caused by the oxidation of NADPH or the increase in A412 caused by the reduction of dithiobis(2-nitrobenzoic acid).
This kit contains reagents for the spectrophotometric determination of glutathione reductase activity either by following the decrease in absorbance caused by the oxidation of NADPH at 340 nm (UV assay) or the increase in absorption caused by the reduction of dithiobis(2-nitrobenzoic acid) (DTNB) at 412 nm (colorimetric assay). This kit provides reagents for a spectrophotometric assay for measuring the activity of glutathione reductase either by following the decrease in A340 caused by the oxidation of NADPH or the increase in A412 caused by the reduction of dithiobis(2-nitrobenzoic acid).
This kit provides reagents for a spectrophotometric assay for measuring the activity of glutathione reductase either by following the decrease in A340 caused by the oxidation of NADPH or the increase in A412 caused by the reduction of dithiobis(2-nitrobenzoic acid).

Analysis Note

UV assay: One unit will cause the oxidation of 1.0 μmole of NADPH at 25 °C at pH 7.5.
Colorimetric assay: One unit will cause the reduction of 1.0 μmole of DTNB to TNB at 25 °C at pH 7.5.

Kit Components Also Available Separately

Product No.
Description
SDS

  • D81305,5′-Dithiobis(2-nitrobenzoic acid) 50 mgSDS

  • G4626Glutathione, oxidized, disodium salt 100 mgSDS

  • N6505β-Nicotinamide adenine dinucleotide 25 mgSDS

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 2

target_organs

Eyes,Central nervous system

wgk

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin R Prigge et al.
Cell reports, 19(13), 2771-2781 (2017-06-29)
Energetic nutrients are oxidized to sustain high intracellular NADPH/NADP+ ratios. NADPH-dependent reduction of thioredoxin-1 (Trx1) disulfide and glutathione disulfide by thioredoxin reductase-1 (TrxR1) and glutathione reductase (Gsr), respectively, fuels antioxidant systems and deoxyribonucleotide synthesis. Mouse livers lacking both TrxR1 and
Glutathione: Chemical, Biochemical and Metabolic Aspects.
Dolphin D., et al.
Glutathione: Chemical, Biochemical and Metabolic Aspects (1989)
I K Smith et al.
Analytical biochemistry, 175(2), 408-413 (1988-12-01)
A method for assaying glutathione reductase (GSH; EC 1.6.4.2) in crude plant extracts is described. The method is based on the increase in absorbance at 412 nm when 5,5'-dithiobis(2-nitrobenzoic acid) (DTNB) is reduced by GSH. The effects of the following
A comparative cytotoxicity study of TiO 2 nanoparticles under light and dark conditions at low exposure concentrations
Dalai S, et al.
Toxicology Research, 1(2), 116-130 (2012)
Karl W Engel et al.
Journal of biophotonics, 9(11-12), 1148-1156 (2016-07-09)
Photobiomodulation (PBM) therapy has been noted to promote cell proliferation and growth in many different cell types shown both in vitro and in vivo. Currently, treatment regimens are used in the clinic for a variety of ailments, including wound healing.

Related Content

Global Trade Item Number

SKUGTIN
GRSA-1KT04061835545421

Questions

1–8 of 8 Questions  
  1. ¿Este KIT sirve para utilizar muestras de suero o plasma en ratas?

    1 answer
    1. This kit should be suitable for use with serum or plasma samples from rats as it is suitable for the measurement of glutathione reductase activity in biological samples.

      Helpful?

  2. In your product bulletin you say you have tested Product GRSA, Glutathione Reductase Assay Kit, on rabbit reticulocyte lysate and Saccharomyces extract. Have you tested it on other species?

    1 answer
    1. At this time, we have used the kit to test only these species. We have received inquiries concerning its use for rat and higher plant tissue, but we have no data at this time.

      Helpful?

  3. How does the storage temperature relate to shipping conditions?

    1 answer
    1. The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.

      Helpful?

  4. Do you know if Product GRSA, Glutathione Reductase Assay Kit, can be used with a 96 well plate reader?

    1 answer
    1. We have used this kit ony with a UV/Vis spectrophotometer with 1 cm pathlength cuvettes. We have not attempted to modify it for use with a plate reader.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Is Product GRSA, Glutathione Reductase Assay Kit, compatible with your CelLytic cell lysis reagents?

    1 answer
    1. We have not tried using this kit with our CelLytic reagents. We believe it should be suitable for use with C2978 for mammalian cell lysis or C4882 for yeast cell lysis.

      Helpful?

  7. In your brochure for Nitric Oxide and Cell Stress you present absorbance data from yeast lysis. How were the yeast prepared?

    1 answer
    1. Here is the procedure as recommended by our Production group (March 2008): Grow S. cerevisiae (we used GP460 private strain) on YEPD medium; harvest cells by centrifugation. Wash the cells [7g]  with 20 ml of 20 mM HEPES buffer, pH 7.0. Discard supernatant. Add 1 ml HEPES buffer to make a thick slurry and pour dropwise into liquid nitrogen. Take approx. 2 g of frozen pellet and homogenize with UltraTurrax T-25 while still in liquid nitrogen to make a fine powder. After all the liquid nitrogen has evaporated, take approximately 2 mL of HEPES buffer and add to the 2 g of finely powdered cells. Centrifuge for 5 min in an Eppendorf (benchtop) centrifuge at maximum speed. The supernatant  liquid is the source of the enzyme. The total amount of protein received from this procedure was 21 mg (8.8 mg/mL in 2.4 mL total volume). We determined an activity of approximately 0.3 units/mL, using both the UV and DTNB assays.

      Helpful?

  8. Is Product GRSA, Glutathione Reductase Assay Kit, suitable for use for GR determination in whole blood?

    1 answer
    1. We do not recommend the kit for use with whole blood due to interference from hemoglobin.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service