Skip to Content
Merck
All Photos(2)

Key Documents

MAK223

Sigma-Aldrich

Aldolase Activity Colorimetric Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

Aldolase Colorimetric Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

cancer; neurological disorders; hematological disorder

storage temp.

−20°C

General description

Aldolase (fructose-bisphosphate aldolase; EC 4.1.2.13) catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three isoforms of aldolase (A, B, and C) are differentially expressed during development. Aldolases have been recognized as positive regulators of the Wnt signaling pathway. Deficiency of aldolases is associated with myopathy and hemolytic anemia.

The Aldolase Activity Colorimetric Assay Kit is a simple and high throughput assay for measuring aldolase activity in serum, plasma, and a variety of tissues and cells. Aldolase activity is determined by measuring a colorimetric product with absorbance at 450 nm (A450) proportional to the enzymatic activity present. One unit of aldolase is the amount of enzyme that generates 1.0 μmole of NADH per minute at pH 7.2 at 37 °C.
Aldolase (fructose-bisphosphate aldolase; EC 4.1.2.13) catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three isoforms of aldolase (A, B, and C) are differentially expressed during development. Aldolases have been recognized as positive regulators of the Wnt signaling pathway. Deficiency of aldolases is associated with myopathy and hemolytic anemia

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of Aldolase activity in biological samples such as tissue, cells, serum, and plasma.

Principle

The Aldolase Activity Colorimetric Assay Kit is a simple and high throughput assay for measuring aldolase activity in serum, plasma, and a variety of tissues and cells. Aldolase activity is determined by measuring a colorimetric product with absorbance at 450 nm (A450) proportional to the enzymatic activity present. One unit of aldolase is the amount of enzyme that generates 1.0 μmole of NADH per minute at pH 7.2 at 37 °C.

Kit Components Only

Product No.
Description

  • Aldolase Assay Buffer

  • Aldolase Substrate

  • Aldolase Enzyme Mix

  • Aldolase Developer

  • NADH Standard

  • Aldolase Positive Control

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldolase positively regulates of the canonical Wnt signaling pathway.
Caspi M, et al.
Molecular Cancer, 13(1), 164-164 (2014)
Zhenchang Guo et al.
Molecular & cellular proteomics : MCP, 20, 100053-100053 (2021-02-10)
Esophageal squamous cell carcinoma (ESCC) is an aggressive malignancy with poor therapeutic outcomes. However, the alterations in proteins and posttranslational modifications (PTMs) leading to the pathogenesis of ESCC remain unclear. Here, we provide the comprehensive characterization of the proteome, phosphorylome
Unraveling the structural and functional features of an aldolase A mutant involved in the hemolytic anemia and severe rhabdomyolysis reported in a child.
Esposito G, et al.
Blood, 105(2), 905-906 (2005)
Human aldolase A natural mutants: relationship between flexibility of the C-terminal region and enzyme function.
Esposito G, et al.
The Biochemical Journal, 380(1), 51-56 (2004)
Biochemical markers of muscular damage.
Brancaccio P, et al
Clinical Chemistry and Laboratory Medicine, 48(6), 757-767 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service