MAK117
Glycerol Assay Kit
sufficient for 200 colorimetric or fluorometric tests
Synonym(s):
Glycerin Assay Kit, Glycerine Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 200 colorimetric or fluorometric tests
application(s)
cosmetics
environmental
food and beverages
detection method
colorimetric
fluorometric
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−20°C
General description
Glycerol, also referred to as glycerin or glycerine, is a 3-carbon sugar alcohol that forms the backbone of fatty acids. It is also a component of phospholipids and triglycerides. Glycerol is a sweet-tasting, hygroscopic liquid. Glycerol is widely used in food, beverage, and pharmaceutical formulations and is the main waste by-product of biodiesel production via transesterification.
Application
Glycerol assay kit has been used to quantify glycerol release from mouse 3T3-L1 cell culture supernatant samples and to measure plasma glycerol levels.
Features and Benefits
Compatible with high-throughput handling systems.
Suitability
This kit is suitable for glycerol detection in foods, beverages, pharmaceutical formulations, and biological samples.
Principle
In this kit, glycerol concentration is determined by a coupled enzyme assay involving glycerol kinase and glycerol phosphate oxidase, resulting in a colorimetric (570 nm)/fluorometric (lex = 535/lem = 587 nm) product, proportional to the glycerol present. The linear range of detection for this kit is 10-1,000 μM (colorimetric) and 2-50 μM (fluorometric).
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Adipose Lipolysis Unchanged by Preexercise Carbohydrate Regardless of Glycemic Index.
Med Sci Sports Exerc., 50(4), 827-836 (2018)
Experimental dermatology, 27(6), 589-602 (2018-04-01)
Dermal white adipose tissue (DWAT) is a main component of human skin, composed of individual lipid-laden mesenchymal cells known as dermal adipocytes (DAs). Besides their well-known role in lipid storage and release, DAs also promote skin immunity, wound healing and
Diabetologia, 62(8), 1463-1477 (2019-05-18)
Chronic glucocorticoid therapy causes insulin resistance, dyslipidaemia, abnormal fat accumulation, loss of muscle mass and osteoporosis. Here we describe a hitherto unknown sexual dimorphism in the metabolic response to chronic glucocorticoid exposure in mice. This led us to investigate whether
Frontiers in physiology, 9, 1452-1452 (2018-11-09)
Background: Impaired capillary growth (angiogenesis) in skeletal muscle and adipose tissue contributes to the development of metabolic disorders in obese males. This association remains unexplored in females, despite mounting evidence that endothelial cells have sex-specific transcriptional profiles. Therefore, herein we
Health & Drugs: Disease, Prescription & Medication (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service