Skip to Content
Merck
All Photos(1)

Documents

MAK266

Sigma-Aldrich

Triglyceride Quantification Colorimetric/Fluorometric Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

TG Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric
fluorometric

relevant disease(s)

neurological disorders; cardiovascular diseases

storage temp.

−20°C

General description

The new Triglyceride Quantification Kit, MAK564, is now available! Triglycerides (TG) are the main constituents of vegetable oil, animal fat, LDL and VLDL, and play an important role as transporters of fatty acids as well as an energy source. TG is broken down into fatty acids and glycerol, after which both can serve as substrates for energy producing and metabolic pathways. High blood levels of TG are implicated in atherosclerosis, heart disease and stroke as well as in pancreatitis.

Suitability

Suitable for use with serum, plasma, and other biological fluids, and tissue and cell culture samples.

Principle

The Triglyceride Quantification Kit provides a sensitive, easy assay to measure TG concentration in a variety of samples. In this assay, TG are converted to free fatty acids and glycerol. The glycerol is then oxidized to generate a colorimetric (570 nm)/fluorometric (lex = 535 nm/lem = 587 nm) product. The kit is sensitive to detect 2 pmole–10 nmole (2–10,000 mM range) of triglyceride in various samples. The kit also detects monoglycerides and diglycerides.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingping Luo et al.
Acta pharmaceutica Sinica. B, 11(3), 668-679 (2021-03-30)
Alcoholic liver disease (ALD) causes insulin resistance, lipid metabolism dysfunction, and inflammation. We investigated the protective effects and direct regulating target of S-allylmercaptocysteine (SAMC) from aged garlic on liver cell injury. A chronic ethanol-fed ALD in vivo model (the NIAAA model)
The role of triglycerides in atherosclerosis.
Talayero, Beatriz G., and Frank M. Sacks.Talayero, Beatriz G and Frank M S
Current Cardiology Reports, 13(6), 544-544 (2011)
David A. Wilson
Clinical Veterinary Advisor: The Horse (2011)
Clinical biochemistry and hematology.
The laboratory rabbit, guinea pig, hamster, and other rodents., 57-116 (2012)
Joseph Pierre Aboumsallem et al.
International journal of molecular sciences, 20(6) (2019-03-16)
The risk of heart failure (HF) is prominently increased in patients with type 2 diabetes mellitus. The objectives of this study were to establish a murine model of diabetic cardiomyopathy induced by feeding a high-sugar/high-fat (HSHF) diet and to evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service