Skip to Content
Merck
All Photos(4)

Documents

MAK071

Sigma-Aldrich

Pyruvate Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Assay Kit for Pyruvate Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

gastrointestinal diseases; cancer

storage temp.

−20°C

Gene Information

General description

Pyruvate is a central molecule in metabolism that is used and synthesized through multiple metabolic pathways including glycolysis and the transamination of alanine. Pyruvate may be further oxidized in the citric acid cycle, converted to carbohydrates during gluconeogenesis, or reduced to lactate. High levels of pyruvate are associated with liver disease and genetic disorders. Pyruvate has also been used to stimulate metabolism leading to loss of body weight.

Suitability

Suitable for measuring pyruvate concentration in various biological samples such as blood, tissues, and culture cells.

Principle

Pyruvate concentration is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the pyruvate present.

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abstracts of the XVIth International Conference on Biological Inorganic Chemistry, July 22-26, 2013, Grenoble, France.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 19 Suppl 1, 1-696 (2014-03-13)
Changes in muscle cell metabolism and mechanotransduction are associated with myopathic phenotype in a mouse model of collagen VI deficiency.
De Palma S, et al.
PLoS ONE, 8(2), e56716-e56716 (2013)
Xiaolong Zhang et al.
Biotechnology and bioengineering, 115(9), 2217-2231 (2018-06-14)
N-acetylneuraminic acid (NeuAc) is widely used as a nutraceutical for facilitating infant brain development, maintaining brain health, and enhancing immunity. Currently, NeuAc is mainly produced by extraction from egg yolk and milk, or via chemical synthesis. However, its low concentration
Lauren K Wareham et al.
Antioxidants & redox signaling, 24(17), 1013-1028 (2016-02-26)
Carbon monoxide is a respiratory poison and gaseous signaling molecule. Although CO-releasing molecules (CORMs) deliver CO with temporal and spatial specificity in mammals, and are proven antimicrobial agents, we do not understand the modes of CO toxicity. Our aim was
Astrocytic metabolic switch is a novel etiology for Cocaine and HIV-1 Tat-mediated neurotoxicity.
Natarajaseenivasan K, et al.
Cell Death & Disease, 9(4), 415-415 (2018)

Articles

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Warburg effect enhances glucose to lactate conversion in tumor cells, regardless of oxygen levels; impacting cancer metabolism since 1924.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service