Skip to Content
Merck
All Photos(1)

Documents

M7278

Sigma-Aldrich

Minimum Essential Medium Eagle

HEPES modification, with Earle′s salts and sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Minimum Essential Medium Eagle, HEPES Modification, with Earle′s salts, 25 mM HEPES and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

>7.2

components

L-glutamine: no
phenol red: yes
HEPES: 25 mM
sodium pyruvate: no
Earle’s salts (5% CO2): yes
NaHCO3: yes

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium Eagle comprises sugar, inorganic salts, water-soluble vitamins and essential amino acids. It is widely used as a media formulation for many cell culture studies.

Application

Minimum Essential Medium Eagle has been used for culturing primordial follicles and ovary tissues, human skin fibroblast cells, human colonic epithelial cell line LS174T and hippocampal tissue slices.

Reconstitution

Supplement with 0.292 g/L L-glutamine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo Lerchundi et al.
Frontiers in cellular neuroscience, 14, 80-80 (2020-05-07)
Genetically encoded nanosensors such as the FRET-based adenosine triphosphate (ATP) sensor ATeam enable the measurement of changes in ATP levels inside cells, promoting our understanding of metabolic interactions between astrocytes and neurons. The sensors are usually well characterized in vitro
Samuel Wilson Failor et al.
Neural development, 13(1), 4-4 (2018-03-27)
Activity in neurons drives afferent competition that is critical for the refinement of nascent neural circuits. In ferrets, when an eye is lost in early development, surviving retinogeniculate afferents from the spared eye spread across the thalamus in a manner
Shafagh A Waters et al.
The EMBO journal, 36(3), 374-387 (2016-11-12)
RNA sequencing studies have identified hundreds of non-coding RNAs in bacteria, including regulatory small RNA (sRNA). However, our understanding of sRNA function has lagged behind their identification due to a lack of tools for the high-throughput analysis of RNA-RNA interactions
Carolina Rodriguez Jimenez et al.
Animal reproduction science, 201, 22-31 (2018-12-21)
Manipulation of oocytes enclosed in preantral follicles (MOEPF) allows for analyzing follicular development and use of this biotechnology in the pre-analysis of the beneficial or toxic effects of bio-products on granulosa cells and oocytes at different developmental stages. In this
Alfred K Yu et al.
Human molecular genetics, 24(10), 2848-2860 (2015-02-06)
Mitochondrial complex I (NADH dehydrogenase) is a major contributor to neuronal energetics, and mutations in complex I lead to vision loss. Functional, neuroanatomical and transcriptional consequences of complex I deficiency were investigated in retinas of the Ndufs4 knockout mouse. Whole-eye

Articles

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

A large selection of MEM formulations. Minimum Essential Medium (MEM), developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service