Skip to Content
Merck
All Photos(2)

Key Documents

M6781

Sigma-Aldrich

Isopalmitic acid

≥98% (capillary GC)

Synonym(s):

14-Methylpentadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H32O2
CAS Number:
Molecular Weight:
256.42
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (capillary GC)

form

powder

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)CCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-15(2)13-11-9-7-5-3-4-6-8-10-12-14-16(17)18/h15H,3-14H2,1-2H3,(H,17,18)

InChI key

ZONJATNKKGGVSU-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Nadathur et al.
Mutation research, 359(3), 179-189 (1996-04-04)
We describe here the isolation and identification of palmitic acid as being responsible for significant anti-N-methyl-N'-nitro-N-nitroguanidine (MNNG) activity in yogurt. The Ames test (Salmonella typhimurium TA100) was used to direct fractionation of activity. Yogurt was freeze-dried and extracted with acetone
I Wollowski et al.
The Journal of nutrition, 129(1), 77-82 (1999-01-23)
Lactic acid-producing bacteria prevent carcinogen-induced preneoplastic lesions and tumors in rat colon. Because the mechanisms responsible for these protective effects are unknown, two strains of lactic acid bacteria, Lactobacillus delbrueckii ssp. bulgaricus 191R and Streptococcus salivarius ssp. thermophilus CH3, that
S R Nadathur et al.
Journal of dairy science, 81(3), 664-671 (1998-05-05)
Reconstituted nonfat dry milk was treated with different amounts of lipase from Pseudomonas fluorescens. Hexane extracts of treated milks were dissolved in dimethylsulfoxide and assayed for antimutagenicity using the Ames test (Salmonella typhimurium TA 100) against N-methyl, N'-nitro, N-nitrosoguanidine. Anti-N-methyl
William F Wood
Journal of chemical ecology, 30(2), 479-482 (2004-04-29)
Using GC-MS analysis, 11 major volatile compounds were found in the preorbital gland secretion from a female sika deer, Cervus nippon. These compounds are the C14 through C18 straight-chain fatty acids, (ZZ)-9,12-octadecadienoic acid, 12-methyltridecanoic acid, 13-methyltetradecanoic acid, 14-methylpentadecanoic acid, 14-methylhexadecanoic
Liang Jie et al.
Food & function, 9(3), 1747-1754 (2018-03-03)
Breast milk consumption reduces the incidence of necrotizing enterocolitis (NEC) in preterm infants compared to formula. Branched-chain fatty acids (BCFAs) are present in breast milk but not in most formulas intended for preterm infants. We aimed to determine the composition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service