Skip to Content
Merck
All Photos(1)

Key Documents

464449

Sigma-Aldrich

2-Hexyldecanoic acid

96%

Synonym(s):

2-n-Hexyldecanoic acid, Isopalmitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH[(CH2)5CH3]CO2H
CAS Number:
Molecular Weight:
256.42
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.446 (lit.)

bp

268 °C (lit.)

mp

18 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCCCCCC(CCCCCC)C(O)=O

InChI

1S/C16H32O2/c1-3-5-7-9-10-12-14-15(16(17)18)13-11-8-6-4-2/h15H,3-14H2,1-2H3,(H,17,18)

InChI key

JMOLZNNXZPAGBH-UHFFFAOYSA-N

General description

2-Hexyldecanoic acid is a carboxylic acid having branched carbon chains.

Application

2-Hexyldecanoic acid may be used as amidating agent for the suppression of side-chain crystallization during the synthesis of amphiphilic macromolecules. It may be employed as a surrogate carboxylic acid in a study.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed H Mohamed et al.
Rapid communications in mass spectrometry : RCM, 23(23), 3703-3712 (2009-11-10)
Aqueous solutions containing simple model aliphatic and alicyclic carboxylic acids (surrogates 1-4) were studied using negative ion electrospray mass spectrometry (ESI-MS) in the presence and absence of alpha-, beta-, and gamma-cyclodextrin. Molecular ions were detected corresponding to the parent carboxylic
Ester synthesis by the lipase from Pseudomonas fragi 22.39 B.
Nishio T, et al.
Agricultural and Biological Chemistry, 52(5), 1203-1208 (1988)
Core-shell-structured highly branched poly (ethylenimine amide) s: synthesis and structure.
Antonietti L, et al.
Macromolecules, 38(14), 5914-5920 (2005)
Shixiong Gao et al.
Environmental science & technology, 53(5), 2862-2872 (2019-02-12)
Exposure to environmental chemicals could disturb lipidome homeostasis in biotas. Comprehensive identification and interpretation of lipid molecules in biological samples are of great importance to elucidate the potential changes in lipid homeostasis upon exposure to various environmental stimuli. In this
Liheng Wu et al.
Nature, 548(7666), 197-201 (2017-08-02)
Crystallization of colloidal nanocrystals into superlattices represents a practical bottom-up process with which to create ordered metamaterials with emergent functionalities. With precise control over the size, shape and composition of individual nanocrystals, various single- and multi-component nanocrystal superlattices have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service