Skip to Content
Merck
All Photos(3)

Key Documents

M5500

Sigma-Aldrich

Melibiose

≥98% (HPLC)

Synonym(s):

α-D-Melibiose hydrate, 6-α-D-Galactopyranosyl-D-glucopyranose, 6-O-α-D-Galactopyranosyl-D-glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

mp

182 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless

SMILES string

O.OC[C@H]1O[C@H](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H22O11.H2O/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10-,11+,12-;/m0./s1

InChI key

CHIDEFLSUMQFBY-CQIZOMOZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Melibiose is a carbon substrate. [1]

Application

Melibiose has been used in a study to improve the preparation of benzoylated ethyl 1-thioglycosides. [2] It has also been used in a study to investigate the effect of water plasticization on the molecular mobility and crystallization tendency of amorphous disaccharides. [3]

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activity of MUC1 cancer antigen-binding plasma anti-α-galactoside antibody correlates inversely with size of autologous lipoprotein(a).
Jessy John et al.
Experimental biology and medicine (Maywood, N.J.), 244(11), 893-900 (2019-08-10)
Parameswaran Hariharan et al.
BMC biology, 16(1), 85-85 (2018-08-05)
Membrane lipids play critical roles in the structure and function of membrane-embedded transporters. Salmonella typhimurium MelB (MelBSt) is a symporter coupling melibiose translocation with a cation (Na+, Li+, or H+). We present an extensive study on the effects of specific
Priya Voothuluru et al.
Frontiers in plant science, 11, 392-392 (2020-05-07)
Reactive oxygen species (ROS) can act as signaling molecules involved in the acclimation of plants to various abiotic and biotic stresses. However, it is not clear how the generalized increases in ROS and downstream signaling events that occur in response
Ville Aumala et al.
ChemSusChem, 12(4), 848-857 (2018-12-28)
Plant-derived carbohydrates are an abundant renewable resource. Transformation of carbohydrates into new products, including amine-functionalized building blocks for biomaterials applications, can lower reliance on fossil resources. Herein, biocatalytic production routes to amino carbohydrates, including oligosaccharides, are demonstrated. In each case
Kieran James et al.
Scientific reports, 9(1), 15427-15427 (2019-10-30)
A number of bifidobacterial species are found at a particularly high prevalence and abundance in faecal samples of healthy breastfed infants, a phenomenon that is believed to be, at least partially, due to the ability of bifidobacteria to metabolize Human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service