Skip to Content
Merck
All Photos(2)

Key Documents

M3650

Sigma-Aldrich

Methyl myristoleate

≥98.5% (capillary GC), liquid

Synonym(s):

Methyl cis-9-tetradecenoate, Myristoleic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
Beilstein:
2046023
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98.5% (capillary GC)

form

liquid

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]\C(CCCC)=C(/[H])CCCCCCCC(=O)OC

InChI

1S/C15H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h6-7H,3-5,8-14H2,1-2H3/b7-6-

InChI key

RWIPSJUSVXDVPB-SREVYHEPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Yi Kong et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(11), 1431-1436 (2006-11-14)
The binary diffusion coefficients, D(12), and retention factors for myristoleic acid and its methyl ester at infinite dilution were measured by the chromatographic impulse response technique in supercritical carbon dioxide at temperatures of 313.2, 333.2 and 343.2 K and pressures
Zhidong Xu et al.
Lipids, 45(2), 199-208 (2010-01-19)
The existing protocols for analyzing fatty acid methyl esters (FAMEs) using a one-step acetyl chloride (AC) catalyzed transesterification and extraction procedure cannot accurately determine the medium- and long-chain fatty acids simultaneously in clinical (enteral, parenteral) formulations. For example: (1) addition
Saskia Thurnhofer et al.
Journal of agricultural and food chemistry, 53(23), 8896-8903 (2005-11-10)
A method using gas chromatography/electron ionization-mass spectrometry (GC/EI-MS) in the selected ion monitoring (SIM) mode was developed for the analysis of fatty acids as methyl esters (FAMEs) in order to determine their percentage contribution to the fatty acid profile in
Tomás Horák et al.
Journal of agricultural and food chemistry, 57(23), 11081-11085 (2009-11-13)
Solid-phase extraction (SPE), solid-phase microextraction (SPME) using carbowax/divinylbenzen fiber, and stir bar sorptive extraction (SBSE) followed by solvent back extraction have been used for the extraction of free fatty acids (caproic, caprylic, pelargonic, capric, lauric, myristic, palmitic, stearic, oleic, linoleic
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service