Skip to Content
Merck
All Photos(2)

Key Documents

A9298

Sigma-Aldrich

Methyl arachidonate

≥99% (GC)

Synonym(s):

Arachidonic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O2
CAS Number:
Molecular Weight:
318.49
Beilstein:
1714445
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

Mortierella alpina

Quality Level

Assay

≥99% (GC)

form

liquid

functional group

ester

lipid type

omega FAs

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC

InChI

1S/C21H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h7-8,10-11,13-14,16-17H,3-6,9,12,15,18-20H2,1-2H3/b8-7-,11-10-,14-13-,17-16-

InChI key

OFIDNKMQBYGNIW-ZKWNWVNESA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Lipid peroxidation products mediate the formation of 8-hydroxydeoxyguanosine in DNA.: This article explores the role of lipid peroxidation products, specifically those involving methyl arachidonate, in causing oxidative DNA damage, indicating the compound′s influence on cellular aging and disease processes (Park JW et al., 1992).

Biochem/physiol Actions

Unlike most unsaturated fatty acid methyl esters, methyl arachidonate is a potent activator of protein kinase C at 5–50 μM. At the low end of the effective concentration range, the effect is due to cyclooxygenase products, while lipoxygenase products mediate the effect at higher concentrations.
Unlike most unsaturated fatty acid methyl esters, methyl arachidonate is a potent activator of protein kinase C.

Packaging

Sealed ampule under vacuum.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjia Tang et al.
Bioresource technology, 196, 559-565 (2015-08-22)
A novel two-step enzymatic method is described in this study to synthesize symmetrical triacylglycerols (TAGs) with arachidonic acid (ARA) at the sn-2 position. The processes included the synthesis of 2-monoacylglycerols (2-MAGs) rich in 2-arachidonoylglycerol (2-AG) by enzymatic ethanolysis and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service