Skip to Content
Merck
All Photos(2)

Key Documents

M2314

Sigma-Aldrich

6-Monodeoxy-6-monoamino-β-cyclodextrin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H71O34N · HCl
CAS Number:
Molecular Weight:
1170.46
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

powder

Quality Level

color

off-white

mp

200-222  °C

solubility

water: soluble

storage temp.

2-8°C

SMILES string

NC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@H]3[C@H](O)[C@@H](O)[C@@H](O[C@H]4[C@H](O)[C@@H](O)[C@@H](O[C@H]5[C@H](O)[C@@H](O)[C@@H](O[C@H]6[C@H](O)[C@@H](O)[C@@H](O7)O[C@@H]6CO)O[C@@H]5CO)O[C@@H]4CO)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O[C

InChI

1S/C43H73NO33.ClH/c44-2-11-32-19(53)25(59)38(66-11)73-33-13(5-47)68-40(27(61)21(33)55)75-35-15(7-49)70-42(29(63)23(35)57)77-37-16(8-50)71-43(30(64)24(37)58)76-36-14(6-48)69-41(28(62)22(36)56)74-34-12(4-46)67-39(26(60)20(34)54)72-31-9(3-45)1-10(65-32)17(51)18(31)52;/h9-43,45-64H,1-8,44H2;1H/t9-,10+,11-,12-,13-,14-,15-,16-,17+,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-;/m1./s1

InChI key

VOAFELNSKVTNFT-BYWRREOMSA-N

Application

6-Monodeoxy-6-monoamino-β-cyclodextrin hydrochloride has been used for capillary electrophoresis. It has been used in a study to demonstrate that covalently attached molecular adapters can alter the ion selectivity of the α-hemolysin pore.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Chen Wu et al.
Journal of the American Chemical Society, 129(51), 16142-16148 (2007-12-01)
Molecular adapters are crucial for the stochastic sensing of organic analytes with alpha-hemolysin (alphaHL) protein nanopores when direct interactions between analytes and the pore cannot readily be arranged by conventional protein engineering. In our earlier studies, cyclodextrin adapters were lodged
Youchang Xiao et al.
Langmuir : the ACS journal of surfaces and colloids, 23(26), 12990-12996 (2007-11-21)
The enhanced enantiomeric separation of racemic phenylalanine solution has been demonstrated by the membrane-based chiral resolution method using an acetylated beta-cyclodextrin-immobilized cellulose dialysis membrane. Beta-cyclodextrin (CD) was first immobilized onto the surface of commercial cellulose dialysis membranes, followed by the
Synthesis and characterization of novel poly(γ-benzyl-L-glutamate) derivatives tailored for the preparation of nanoparticles of pharmaceutical interest
Martinez Barbosa, M., et al.
Polymer International, 56, 317-324 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service