Skip to Content
Merck
All Photos(3)

Key Documents

C4767

Sigma-Aldrich

β-Cyclodextrin

≥97%

Synonym(s):

beta-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
Beilstein:
78623
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

biological source

microbial

Quality Level

Assay

≥97%

form

powder

optical activity

[α]20/D +162±3°, c = 1.5% in H2O

mp

290-300 °C (dec.) (lit.)

solubility

1 M NH4OH: 50 mg/mL

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Cyclodextrin is a cyclic oligosaccharide consisting of seven glucose subunits joined by α-(1,4) glycosidic bonds forming a truncated conical structure. It is widely used in food, pharmaceutical, cosmetics, and chemicals industries.

Application

β-Cyclodextrin is used as a complexing agent in drug delivery because it forms an inclusion complex with a drug molecule. The complex of cyclodextrin increases the aqueous solubility, dissolution rate and bioavailability of poorly water-soluble drugs, which is useful for the delivery of the medical agent to a biological system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of dehydrated beta-cyclodextrin as pharmaceutical excipient
Martini A, et al.
Drug Development and Industrial Pharmacy, 20, 2381-2393 (1994)
Preparation and characterization of microparticles of ?-cyclodextrin/glutathione and chitosan/glutathione obtained by spray-drying
Webber V, et al.
Food Research International, 105, 432-439 (2018)
Study to explore the mechanism to form inclusion complexes of beta-cyclodextrin with vitamin molecules
Saha S, et al.
Scientific Reports, 6, 35764-35764 (2016)
Inclusion complexation of tolbutamide with ?-cyclodextrin and hydroxypropyl-?-cyclodextrin
Veiga F, et al.
Journal of psychosomatic obstetrics and gynaecology, 129, 63-71 (1996)
Cyclodextrins
Kurkov SV and Loftsson T
The International journal of pharmacy practice, 453, 167-180 (2013)

Articles

Techniques for solubilizing metabolically important compounds in aqueous solutions are reviewed.

Techniques for solubilizing metabolically important compounds in aqueous solutions are reviewed.

Techniques for solubilizing metabolically important compounds in aqueous solutions are reviewed.

Techniques for solubilizing metabolically important compounds in aqueous solutions are reviewed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service