Skip to Content
Merck
All Photos(1)

Key Documents

L9381

Sigma-Aldrich

Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia)

FITC conjugate, lyophilized powder

Synonym(s):

Bandeirea simplicifolia agglutinin, BS-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

Bandieraea simplicifolia

Quality Level

conjugate

FITC conjugate

form

lyophilized powder

potency

<100 μg per mL agglutination activity (using human blood group A erythrocytes)

composition

Protein, ~10% Lowry

extent of labeling

1-3 mol FITC per mol protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

BS-I has a major affinity for terminal α-D-galactosyl residues with a secondary affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Other Notes

BS-I is a tetrameric lectin consisting of two types of subunits designated A and B. There are five BS-I isolectins with different subunit composition: BSI-B4, BSI-AB3, BSI-A2B2, BSI-A3B and BSI-A4. BSI-B4 is blood group B specific and has an exclusive affinity for terminal α-D-galactosyl residues, whereas BSI-A4 has blood group A specificity and has a major affinity for terminal N-acetyl-α-D-galactosaminyl residues.

Physical form

Contains phosphate buffer salts and sodium chloride

Analysis Note

Agglutination activity is expressed in μg/mL and is determined from serial dilutions of a 1 mg/mL solution using phosphate buffered saline, pH 6.8, containing, for each lectin, calcium, magnesium, and manganese at different concentrations. This activity is the lowest concentration to agglutinate a 2% suspension of appropriate erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Right Ventricle Vasculature in Human Pulmonary Hypertension Assessed by Stereology.
Brian B Graham et al.
American journal of respiratory and critical care medicine, 196(8), 1075-1077 (2017-03-10)
Jing Feng et al.
Investigative ophthalmology & visual science, 61(8), 47-47 (2020-07-31)
The aim of this study was to investigate apelin and its potential neovascularization role in retinopathy of prematurity (ROP) along with the inhibitory effects of its antagonist. We used an oxygen-induced retinopathy (OIR) mouse model to explore the progress of
Francesco Di Maggio et al.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.], 16(6), 995-1004 (2016-06-12)
The vascular heterogeneity of pancreatic ductal adenocarcinoma (PDAC) has never been characterised. We analysed the heterogeneous vascular density of human PDAC along with its prognostic correlation. Tissue Microarrays of 87 patients with different pancreatico-biliary pathologies were analysed in an automated
David Gau et al.
The Journal of biological chemistry, 295(28), 9618-9629 (2020-05-24)
Angiogenesis-mediated neovascularization in the eye is usually associated with visual complications. Pathological angiogenesis is particularly prominent in the retina in the settings of proliferative diabetic retinopathy, in which it can lead to permanent loss of vision. In this study, by
Mahak Singhal et al.
EMBO molecular medicine, 12(6), e11164-e11164 (2020-04-18)
The angiopoietin (Ang)-Tie pathway has been intensely pursued as candidate second-generation anti-angiogenic target. While much of the translational work has focused on the ligand Ang2, the clinical efficacy of Ang2-targeting drugs is limited and failed to improve patient survival. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service