Skip to Content
Merck
All Photos(1)

Documents

L9006

Sigma-Aldrich

Lectin from Ulex europaeus (gorse, furze)

FITC conjugate, lyophilized powder

Synonym(s):

Ulex europaeus agglutinin, UEA-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

conjugate

FITC conjugate

Quality Level

form

lyophilized powder

potency

<5 μg/mL agglutination activity

composition

Protein, ~10% Lowry

extent of labeling

2-5 mol FITC per mol protein

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ulex europaeus agglutinin has anti-H blood group specificity. There are two types of lectin: UEA I has an affinity for L-fucose and UEA II has an affinity for N,N′-diacetylchitobiose. The mol. wt. of UEA I was initially found to be 170,000 daltons. Later reports indicate that UEA I may form aggregates at neutral and basic pH and that the correct mol. wt. is 68,000 daltons.

Packaging

Package size based on protein content.

Linkage

Conjugates are prepared from affinity purified lectin.

Physical form

Contains potassium phosphate buffer salts and NaCl

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hour incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoki Akanuma et al.
Pancreas, 46(9), 1202-1207 (2017-09-14)
We aimed to evaluate the contribution of acinar-to-ductal metaplasia (ADM) to the accumulation of cells with a ductal phenotype in cultured human exocrine pancreatic tissues and reveal the underlying mechanism. We sorted and cultured viable cell populations in human exocrine
Karla Lehle et al.
Artificial organs, 40(6), 577-585 (2015-10-30)
Multipotent progenitor cells were mobilized during pediatric extracorporeal membrane oxygenation (ECMO). We hypothesize that these cells also adhered onto polymethylpentene (PMP) fibers within the membrane oxygenator (MO) during adult ECMO support. Mononuclear cells were removed from the surface of explanted
Alan Pestronk et al.
Muscle & nerve, 42(1), 53-61 (2010-06-15)
The causes of perifascicular myofiber atrophy and capillary pathology in dermatomyositis are incompletely understood. We studied 11 dermatomyositis muscles by histochemistry, immunohistochemistry, and ultrastructure. We found that endomysial capillaries within regions of perifascicular atrophy are not entirely lost, but they
Zoe E Clayton et al.
International journal of cardiology, 234, 81-89 (2017-02-18)
Endothelial cells derived from human induced pluripotent stem cells (iPSC-ECs) promote angiogenesis, and more recently induced endothelial cells (iECs) have been generated via fibroblast trans-differentiation. These cell types have potential as treatments for peripheral arterial disease (PAD). However, it is
Gang Chen et al.
The Journal of clinical investigation, 119(10), 2914-2924 (2009-09-18)
Various acute and chronic inflammatory stimuli increase the number and activity of pulmonary mucus-producing goblet cells, and goblet cell hyperplasia and excess mucus production are central to the pathogenesis of chronic pulmonary diseases. However, little is known about the transcriptional

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service