Skip to Content
Merck
All Photos(1)

Key Documents

L5288

Sigma-Aldrich

Latrunculin B

from Latruncula magnifica, ≥80% (HPLC), solid, actin polymerization inhibitor

Synonym(s):

LAT-B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29NSO5
CAS Number:
Molecular Weight:
395.51
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Latrunculin B from Latruncula magnifica, ≥80% (HPLC), solid

Quality Level

Assay

≥80% (HPLC)

form

solid

mol wt

395.5

solubility

ethanol: soluble

storage temp.

−20°C

SMILES string

C[C@H]1CC[C@@H]2C[C@H](C[C@@](O)(O2)[C@@H]3CSC(=O)N3)OC(=O)C=C(C)CCC=C1

InChI

1S/C20H29NO5S/c1-13-5-3-4-6-14(2)9-18(22)25-16-10-15(8-7-13)26-20(24,11-16)17-12-27-19(23)21-17/h3,5,9,13,15-17,24H,4,6-8,10-12H2,1-2H3,(H,21,23)/b5-3-,14-9-/t13-,15-,16-,17+,20-/m1/s1

InChI key

NSHPHXHGRHSMIK-JRIKCGFMSA-N

General description

Latrunculin B is a macrocyclic toxin, which is produced by Latruncula magnifica.

Application

Latrunculin B from Latruncula magnifica has been used for actin depolymerisation in various experiments. It has also been used as a component in bone marrow-derived dendritic cell (BMDC) complete medium, to study the formation of tubules from phagosomes in dendritic cells.

Biochem/physiol Actions

Latrunculin B inhibits actin polymerization in vitro. Latrunculin B disrupts microfilament-mediated processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of the theta comparative cell scoring method to quantify diverse phenotypic responses between distinct cell types
Warchal SJ, et al.
Assay and Drug Development Technologies, 14(7), 395-406 (2016)
Microtubule-dependent targeting of the exocyst complex is necessary for xylem development in Arabidopsis
Vukavsinovic N, et al.
The New phytologist, 213(3), 1052-1067 (2017)
Visualizing toll-like receptor-dependent phagosomal dynamics in murine dendritic cells using live cell microscopy
Mantegazza AR and Marks MS
Methods in Molecular Biology, 191-203 (2015)
Synthesis of Essential Drugs (2012)
Steven L Jones et al.
The Journal of cell biology, 205(1), 67-81 (2014-04-09)
The axon initial segment (AIS) of differentiated neurons regulates action potential initiation and axon-dendritic polarity. The latter function depends on actin dynamics, but actin structure and functions at the AIS remain unclear. Using platinum replica electron microscopy (PREM), we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service