Skip to Content
Merck
All Photos(1)

Key Documents

428020

Sigma-Aldrich

Latrunculin B

from Latrunculia magnifica, ≥95% (HPLC), solid, actin polymerization inhibitor, Calbiochem®

Synonym(s):

Latrunculin B, Latrunculia magnifica

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29NO5S
CAS Number:
Molecular Weight:
395.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Latrunculin B, Latrunculia magnifica, Latrunculin B, CAS 76343-94-7, is a unique marine toxin that inhibits actin polymerization and disrupts microfilament organization. It is 10 to 100-fold more potent than cytochalasins.

Quality Level

description

Merck USA index - 14, 5378

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white to yellow

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL
methanol: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

S1C[C@H](NC1=O)[C@@]2(O[C@H]3C[C@@H](OC(=O)\C=C(/CC\C=C/[C@H](CC3)C)\C)C2)O

InChI

1S/C20H29NO5S/c1-13-5-3-4-6-14(2)9-18(22)25-16-10-15(8-7-13)26-20(24,11-16)17-12-27-19(23)21-17/h3,5,9,13,15-17,24H,4,6-8,10-12H2,1-2H3,(H,21,23)/b5-3-,14-9-/t13-,15-,16-,17+,20-/m1/s1

InChI key

NSHPHXHGRHSMIK-JRIKCGFMSA-N

General description

A structurally unique marine toxin. Inhibits actin polymerization in vitro and disrupts microfilament organization as well as microfilament-mediated processes. 10 to 100-fold more potent than cytochalasins.
A structurally unique marine toxin. Inhibits actin polymerization in vitro and disrupts microfilament organization as well as microfilament-mediated processes; 10 to 100-fold more potent than cytochalasins.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Actin polymerization
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Other Notes

Spector, I., et al. 1989. Cell Motil. Cytoskeleton13, 127.
de Oliveira, C.A., and Mantovani, B. 1988. Life Sci.43, 1825.
Coue, M., et al. 1987. FEBS Lett.213, 316.
Spector, I., et al. 1983. Science219, 493.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanumanthu Bd Prasada Rao et al.
eLife, 10 (2021-09-30)
During meiosis, protein ensembles in the nuclear envelope (NE) containing SUN- and KASH-domain proteins, called linker nucleocytoskeleton and cytoskeleton (LINC) complex, promote the chromosome motion. Yeast SUN-domain protein, Mps3, forms multiple meiosis-specific ensembles on NE, which show dynamic localisation for
Giovanni Stefano et al.
Methods in molecular biology (Clifton, N.J.), 2177, 23-33 (2020-07-08)
The endoplasmic reticulum (ER) is one of the most abundant endomembrane compartments and is in close association with most of the other organelles. In mammalian and yeast cells, the physiological roles and the molecular machineries underlying such association have only
Antonios Georgantzoglou et al.
The Journal of cell biology, 221(8) (2022-06-23)
Migrating cells must interpret chemical gradients to guide themselves within tissues. A long-held principle is that gradients guide cells via reorientation of leading-edge protrusions. However, recent evidence indicates that protrusions can be dispensable for locomotion in some contexts, raising questions
Leila Taher et al.
Scientific reports, 11(1), 23731-23731 (2021-12-11)
Superovulation is the epitome for generating oocytes for molecular embryology in mice, and it is used to model medically assisted reproduction in humans. However, whether a superovulated oocyte is normal, is an open question. This study establishes for the first
Kathleen Scheffler et al.
Journal of cell science, 135(6) (2022-02-04)
Formation of healthy mammalian eggs from oocytes requires specialised F-actin structures. F-actin disruption produces aneuploid eggs, which are a leading cause of human embryo deaths, genetic disorders and infertility. We found that oocytes contain prominent nuclear F-actin structures that are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service