Skip to Content
Merck
All Photos(1)

Key Documents

62230

Supelco

Linoleic acid

analytical standard

Synonym(s):

cis-9,cis-12-Octadecadienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
280.45
Beilstein:
1727101
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.466 (lit.)
n20/D 1.470

bp

229-230 °C/16 mmHg (lit.)

mp

−5 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

carboxylic acid

storage temp.

2-8°C

SMILES string

OC(CCCCCCC/C=C\C/C=C\CCCCC)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-7,9-10H,2-5,8,11-17H2,1H3,(H,19,20)/b7-6-,10-9-

InChI key

OYHQOLUKZRVURQ-HZJYTTRNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Linoleic acid is an octadecadienoic acid and has two double bonds at carbon 9 and 12. It is an essential polyunsaturated fatty acid (PUFA) abundantly found in plant oils.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

This analytical standard can also be used as follows:

  • Multi-residue analysis of oleic acid and other related fatty acids using a derivatization-free method based on gas chromatography-flame ionization detection (GC-FID)
  • Separation and quantification of palmitic, oleic, linoleic, and linolenic acids from 186 samples of sea buckthorn seed oil using near-infrared spectroscopy (NIR) in combination with four different combinations of multivariate calibration methods— partial least squares (PLS), PLS-uninformative variables elimination (UVE), PLS- competitive adaptive reweighted sampling (CARS), and multiple linear regression (MLR)-UVE
  • Separation and determination of fatty acids by gas chromatography (GC) and amino acids by ultra-fast liquid chromatography-ultraviolet (UFLC-UV) detection from goldband goatfish harvested during different seasons
  • Ultra-performance liquid chromatography-mass spectrometry (UHPLC) method-based separation, identification, and determination of eight saturated and unsaturated fatty acids in eight different commercial samples of olive oil without any prior sample treatment
  • Magnetic solid-phase extraction of free fatty acids from edible oil samples using monodisperse magnetic single-crystal ferrite (Fe3O4) nanoparticles for their determination by gas chromatography-flame ionization detection (GC-FID)

Biochem/physiol Actions

Linoleic acid increases cell proliferation and gene expression of PPARα and its target genes such as acyl-CoA oxidase in primary duck hepatocytes [1].

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiufen Zhang et al.
Frontiers in physiology, 13, 844148-844148 (2022-03-11)
In the present study, developmental changes of gluconeogenesis and glycolysis in an avian model were measured, and then the intervention effects of in ovo feeding (IOF) linoleic acid (LA) on hepatic glucose metabolism were evaluated. In Experiment 1, thirty fertilized
Sisi Zhang et al.
mAbs, 15(1), 2232486-2232486 (2023-07-07)
Polysorbates (PSs) are a class of surfactants commonly used in the formulation of protein therapeutic agents to provide protection against denaturation and aggregation. When the PS in these drug formulations degrades, loss of stabilization of the protein therapeutic and formulation
F Valianpour et al.
Journal of lipid research, 44(3), 560-566 (2003-02-04)
The object of this study was to investigate whether the levels of cardiolipin in cultured skin fibroblasts of patients with Barth syndrome (BTHS) can be restored by addition of linoleic acid to growth media. To this end, fibroblasts from controls
Antonius H M Terpstra
The American journal of clinical nutrition, 79(3), 352-361 (2004-02-27)
Studies in mice have indicated that feeding diets containing 0.5-1% conjugated linoleic acid (CLA) considerably reduces body fat. These findings have attracted much interest because of the potential use of CLA as a tool to promote weight loss in humans.
Leanne Hodson et al.
The Journal of nutrition, 144(1), 33-41 (2013-11-15)
Erythrocytes, compared with plasma, are considered more robust markers of n-3 (ω-3) polyunsaturated fatty acid (PUFA) intake, because dietary-induced change in fatty acid (FA) composition takes longer to complete. The extent to which this applies to intakes of saturated fatty

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service