Skip to Content
Merck
All Photos(1)

Key Documents

L1378

Sigma-Aldrich

L-Lactic Dehydrogenase from bovine muscle

Type X, ammonium sulfate suspension, ≥600 units/mg protein

Synonym(s):

(S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD, Lactate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine muscle

type

Type X

Assay

3.0—25.0 mg protein/mL (biuret)

form

ammonium sulfate suspension

specific activity

≥600 units/mg protein

manufacturer/tradename

Sigma-Aldrich

technique(s)

activity assay: suitable

color

white

pH

7.5

UniProt accession no.

application(s)

life science and biopharma

foreign activity

pyruvate kinase ≤0.01%

storage temp.

2-8°C

Gene Information

Related Categories

General description

Research Area: Cell Signaling
Lactate dehydrogenase (LDH) is a key enzyme in the anaerobic metabolic pathway and falls under the class of oxidoreductases. This enzyme is found in a wide range of organisms, including both plants and animals. It is widely distributed across all tissues. LDH is primarily a cytoplasmic enzyme, with particularly high concentrations in muscle, liver, and kidney tissues.

Application

L-Lactic Dehydrogenase from bovine muscle has been used in lactate assay to assess lactate tissue accumulation.

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.
Lactate dehydrogenase (LDH) plays a critical role as a checkpoint in gluconeogenesis and DNA metabolism. It plays a vital role in maintaining homeostasis during periods of low oxygen availability. As an oxidoreductase enzyme, lactate dehydrogenase catalyzes the reversible conversion of pyruvate to lactate using NADH. This enzyme is crucial for the anaerobic metabolism of glucose when oxygen is either absent or in short supply. LDH is the sole serum biomarker effective for evaluating metastatic melanomas.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Crystalline suspension in 2.4 M (NH4)2SO4 solution, pH 6.0

Analysis Note

Protein determined by biuret.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolic recovery from submaximal exercise in hypoxia acclimated high altitude deer mice (Peromyscus maniculatus)
Dessureault LM, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 274, 111004-111004 (2024)
Biochemistry, lactate dehydrogenase
Farhana A and Lappin SL
StatPearls [Internet] (2020)
O V Basharina et al.
Radiatsionnaia biologiia, radioecologiia, 52(6), 602-607 (2013-03-23)
The influence of UV-light (240-390 nm) at the dozes of 151 and 755 J/m2 on the intensity of processes of the lipid peroxidation, activity of lactate dehydrogenase (LDH), succinate dehydrogenase (SDH), cytochrome c oxidase and the level of the energy
Wei Quan et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 33(1), 85-91 (2013-04-20)
To determine the cardioprotective effect of magnesium lithospermate B (MLB) on myocardial ischemia/reperfusion (MI/R) injury and to investigate the antioxidant potential in vivo and in vitro. MI/R injury was induced by the occlusion of left anterior descending coronary artery for
Vasco Meneghini et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(14), 6047-6059 (2013-04-05)
Dysregulated hippocampal neurogenesis has been associated with neurodegenerative disorders, including Alzheimer's disease (AD), in which it may potentially represent an auto-reparatory mechanism that could counteract neuronal loss and cognitive impairment. We evaluated hippocampal neurogenesis in TgCRND8 mice and reported that

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service