Skip to Content
Merck
All Photos(1)

Key Documents

K2389

Sigma-Aldrich

Kainic acid monohydrate

≥98% (HPLC), from Digenea simplex

Synonym(s):

2-Carboxy-3-carboxymethyl-4-isopropenylpyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4 · H2O
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

Digenea simplex

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

O.CC(=C)[C@H]1CN[C@@H]([C@H]1CC(O)=O)C(O)=O

InChI

1S/C10H15NO4.H2O/c1-5(2)7-4-11-9(10(14)15)6(7)3-8(12)13;/h6-7,9,11H,1,3-4H2,2H3,(H,12,13)(H,14,15);1H2/t6-,7+,9-;/m0./s1

InChI key

FZNZRJRSYLQHLT-SLGZUKMRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Kainic acid monohydrate has been used:
  • As a convulsant to induce epileptogenesis and epilepsy in mice.
  • To stimulate in vitro excitotoxic trauma in spiral ganglion neurons on inner hair cells.
  • To induce seizures in rat model.
  • To induce status epilepticus in adult male Wistar rats.

Biochem/physiol Actions

Ionotropic glutamate receptors form ion channels, and conduct Na+ and K+ fluxes. The receptors possess an agonist binding site and it encounters a conformational change upon agonist binding to it. Kainate gated channels participate in glutamate-induced excitatory postsynaptic neuronal potential.
Kainic acid monohydrate is an agonist at the kainate class of ionotropic glutamate receptors, which induces seizures and neurodegeneration in vivo and is used to induce experimental epilepsy in rodents and study the mechanisms of excitation-induced neuronal apoptosis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urokinase-type plasminogen activator deficiency has little effect on seizure susceptibility and acquired epilepsy phenotype but reduces spontaneous exploration in mice
Rantala J, et al.
Epilepsy & Behavior : E&B, 42, 117-128 (2015)
Adeno-associated viral vector-induced overexpression of neuropeptide Y Y2 receptors in the hippocampus suppresses seizures
Woldbye David PD, et al.
Brain, 133(9), 2778-2788 (2010)
Combined gene overexpression of neuropeptide Y and its receptor Y5 in the hippocampus suppresses seizures
Gotzsche CR, et al.
Neurobiology of Disease, 45(1), 288-296 (2012)
Isabel San Martín Molina et al.
Frontiers in neuroscience, 16, 944432-944432 (2022-08-16)
Non-invasive magnetic resonance imaging (MRI) methods have proved useful in the diagnosis and prognosis of neurodegenerative diseases. However, the interpretation of imaging outcomes in terms of tissue pathology is still challenging. This study goes beyond the current interpretation of in
Antti M Airaksinen et al.
Epilepsia, 53(7), 1245-1253 (2012-06-14)
To investigate how kainic acid-induced epileptiform activity is related to hemodynamic changes probed by blood oxygenation level-dependent functional magnetic resonance imaging (BOLD fMRI). Epileptiform activity was induced with kainic acid (KA) (10 mg/kg, i.p.), and simultaneous fMRI at 7 Tesla

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service