Skip to Content
Merck
All Photos(4)

Key Documents

K0250

Sigma-Aldrich

Kainic acid monohydrate

≥99% (TLC), powder, ionotropic glutamate receptor (kainate class) agonist

Synonym(s):

Digenin, Kainate, 2-Carboxy-3-carboxymethyl-4-isopropenylpyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO4 · H2O
CAS Number:
Molecular Weight:
231.25
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Product Name

Kainic acid monohydrate, ≥99% (TLC)

Quality Level

Assay

≥99% (TLC)

form

powder

impurities

Glutamate, free

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

O.CC(=C)[C@H]1CN[C@@H]([C@H]1CC(O)=O)C(O)=O

InChI

1S/C10H15NO4.H2O/c1-5(2)7-4-11-9(10(14)15)6(7)3-8(12)13;/h6-7,9,11H,1,3-4H2,2H3,(H,12,13)(H,14,15);1H2/t6-,7+,9-;/m0./s1

InChI key

FZNZRJRSYLQHLT-SLGZUKMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kainic acid monohydrate serves as an agonist for kainate-class ionotropic glutamate receptors, initiating seizures and neurodegeneration in live organisms. It is employed to induce experimental epilepsy in rodents and to explore the mechanisms underlying excitation-induced neuronal apoptosis.

Application

Kainic acid (KA) has been used:
  • tostudy mechanisms of excitation-induced apoptosis and epilepsy.
  • to hamper themitochondrial function ()
  • used to induce c-fosexpression in the mice′s brains, specifically targeting the dorsal hippocampus.()

Biochem/physiol Actions

Agonist for kainate class of ionotropic glutamate receptors.
Kainic acid monohydrate disrupts mitochondrial function by inducing the release of lactate dehydrogenase (LDH) and reducing 3-(4,5-dimethylthiazole-2-yl)-2,5-diphenyl tetrazolium bromide (MTT). It also triggers the generation of lipid peroxides, which are key mediators in initiating and regulating inflammation and oxidative stress.
Kainic acid monohydrate is an agonist at the kainate class of ionotropic glutamate receptors, which induces seizures and neurodegeneration in vivo and is used to induce experimental epilepsy in rodents and study the mechanisms of excitation-induced neuronal apoptosis.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Excitatory Amino Acid Transporters and Glutamate Receptors (Ion Channel Family) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Dissolve in 1-2 drops of 1N NaOH then bring to volume with water or aqueous buffer. Can be stored 1-2 days refrigerated.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lily M Y Yu et al.
eNeuro, 3(1) (2016-03-30)
Epilepsy is a neurological disorder defined by the presence of seizure activity, manifest both behaviorally and as abnormal activity in neuronal networks. An established model to study the disorder in rodents is the systemic injection of kainic acid, an excitatory
Fudong Liu et al.
Stroke, 40(5), 1842-1848 (2009-03-07)
Over the past 5 years, experimental data have emerged that ischemia-induced cell death pathways may differ in males and females. Cell death in males is triggered by poly(ADP-ribose) polymerase activation and nuclear translocation of apoptosis-inducing factor. We have previously shown
J T Coyle
Ciba Foundation symposium, 126, 186-203 (1987-01-01)
Kainic acid, an acidic pyrolidine isolated from the seaweed Digenea simplex, is the most potent of the commonly used exogenous excitotoxins. The neurotoxic threshold of kainic acid is nearly two magnitudes lower than that of the other receptor-specific agonists, N-methyl-D-aspartic
I Ferrer et al.
Acta neuropathologica, 99(3), 245-256 (2000-02-09)
Caspases play crucial roles in the inflammatory response and in the cell pathway leading to apoptosis. Caspase 1 (ICE), 2 (Nedd2), 3 (CPP32), 6 (Mch2) and 8 (Mch5, FLICE) expression was examined using immunohistochemistry in the brains of rats and
Qun Wang et al.
Molecular neurobiology, 31(1-3), 3-16 (2005-06-15)
Neuronal excitation involving the excitatory glutamate receptors is recognized as an important underlying mechanism in neurodegenerative disorders. Excitation resulting from stimulation of the ionotropic glutamate receptors is known to cause the increase in intracellular calcium and trigger calcium-dependent pathways that

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service