Skip to Content
Merck
All Photos(2)

Key Documents

I5159

Sigma-Aldrich

Imiquimod

≥98% (HPLC), solid, caspase 3 activator

Synonym(s):

1-(2-Methylpropyl)-1H-imidazole[4,5-c]quinoline-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N4
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Imiquimod, ≥98% (HPLC), solid

Assay

≥98% (HPLC)

form

solid

solubility

H2O: <2 mg/mL
DMSO: 4 mg/mL (warming to 60 °C for 15 minutes)

SMILES string

CC(C)Cn1cnc2c(N)nc3ccccc3c12

InChI

1S/C14H16N4/c1-9(2)7-18-8-16-12-13(18)10-5-3-4-6-11(10)17-14(12)15/h3-6,8-9H,7H2,1-2H3,(H2,15,17)

InChI key

DOUYETYNHWVLEO-UHFFFAOYSA-N

Gene Information

human ... TLR7(51284)

Looking for similar products? Visit Product Comparison Guide

General description

Imiquimod is a toll-like receptor (TLR)-7/8 agonist.

Application

Imiquimod has been used as TLR7 agonist for TLR7 agonist enhancement studies. It has also been used as an NLR family, pyrin domain containing 3 (NLRP3) stimulant to treat NLRP3-FL-Btn and FLAG-ASC-FL.

Biochem/physiol Actions

Imiquimod helps to induce the innate cutaneous immunity and the cellular arm of the adaptive immune response.
Imiquimod is a caspase 3 activator, which directly induces procaspase 3 cleavage to active caspase 3. Imiquimod induces apoptosis in vivo in basal cell carcinoma. Its anti-tumor activity is related to the induction of apoptosis. Imiquimod has anti-angiogenic, anti-inflammatory, anti-viral activities. Imiquimod also acts as an immune response modulator inducing the secretion of various cytokines and chemokines.
Imiquimod is a caspase 3 activator, which directly induces procaspase 3 cleavage to active caspase 3. Imiquimod induces apoptosis in vivo and acts as an immunomodulator with anti-viral and anti-inflammatory perties.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topical imiquimod is and effective and safe drug for molluscum contagiosum in children
Badavanis G, et al.
Acta Dermatovenerologica Croatica, 25(2), 164-164 (2017)
Microglial-derived miRNA let-7 and HMGB1 contribute to ethanol-induced neurotoxicity via TLR7
Coleman L G, et al.
Journal of Neuroinflammation, 14(1), 22-22 (2017)
Aimée H M M Arits et al.
The Lancet. Oncology, 14(7), 647-654 (2013-05-21)
Superficial basal-cell carcinoma is most commonly treated with topical non-surgical treatments, such as photodynamic therapy or topical creams. Photodynamic therapy is considered the preferable treatment, although this has not been previously tested in a randomised control trial. We assessed the
Tzu-Kai Lin et al.
Environmental toxicology, 39(9), 4360-4371 (2024-05-18)
The primary function of the skin is to form a mechanical, permeability, antimicrobial, and ultraviolet radiation barrier, which is essential for maintaining physiological homeostasis. Our previous studies demonstrated that cutaneous pigmentation could promote skin barrier function in addition to providing
Juandy Jo et al.
PLoS pathogens, 10(6), e1004210-e1004210 (2014-06-27)
The ability of innate immune cells to sense and respond to impending danger varies by anatomical location. The liver is considered tolerogenic but is still capable of mounting a successful immune response to clear various infections. To understand whether hepatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service