Skip to Content
Merck
All Photos(1)

Key Documents

87917

Sigma-Aldrich

Tetramethylrhodamine ethyl ester perchlorate

suitable for fluorescence, ≥90% (HPCE)

Synonym(s):

TMRE, TMRE perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H27ClN2O7
CAS Number:
Molecular Weight:
514.95
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

Assay

≥90% (HPCE)

form

solid

solubility

DMSO: soluble
alcohols: soluble

fluorescence

λex 540 nm; λem 595 nm in DMSO

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.CCOC(=O)c1ccccc1-c2c3ccc(cc3[o+]c4cc(ccc24)N(C)C)N(C)C

InChI

1S/C26H27N2O3.ClHO4/c1-6-30-26(29)20-10-8-7-9-19(20)25-21-13-11-17(27(2)3)15-23(21)31-24-16-18(28(4)5)12-14-22(24)25;2-1(3,4)5/h7-16H,6H2,1-5H3;(H,2,3,4,5)/q+1;/p-1

InChI key

NBAOBNBFGNQAEJ-UHFFFAOYSA-M

General description

Tetramethylrhodamine ethyl ester perchlorate (TMRE), also known as TMRE perchlorate, is a potential mitochondrial dye. TMRE staining process is reversible, and it doesn’t affect cell proliferation and viability.

The sorted cells containing TMRE perchlorate don’t interfere with the subsequent functional assays and hence are a preferred choice for the enrichment of functionally active, unbiased cell populations.

Application

Tetramethylrhodamine ethyl ester perchlorate (TMRE) is used to study the effects of high glucose in inducing mitochondrial morphology and metabolic changes.

TMRE perchlorate is also used to study Fullerene C 60 penetration into Leukemic Cells and the subsequent photoinduced Cytotoxic Effects.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Potential-sensitive probe for measuring membrane potential changes in mitochondria

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Shan Chen et al.
Cancer management and research, 11, 7353-7367 (2019-09-10)
The role of endothelin receptor type B (EDNRB) isoform 3 involved in Temozolomide (TMZ)-induced melanoma cell death has not yet been elucidated. The subcellular localization of EDNRB isoform 3 was determined by confocal and immunoblotting assays. Silencing EDNRB isoform 3
Natasha S Barteneva et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 62(4), 265-275 (2014-01-08)
High-speed fluorescence-activated cell sorting is relevant for a plethora of applications, such as PCR-based techniques, microarrays, cloning, and propagation of selected cell populations. We suggest a simple cell-sorting technique to eliminate early and late apoptotic and necrotic cells, with good
Garima Sharma et al.
Journal of biochemical and molecular toxicology, 33(8), e22337-e22337 (2019-04-09)
Elevated levels of saturated fatty acids show a strong cytotoxic effect in liver cells. Sirtuin 3 (SIRT3), a mitochondrially localized member of NAD+ -dependent deacetylase has been shown to protect hepatocytes against the oxidative stress. The role of SIRT3 on
Phong Lu et al.
Molecular pharmaceutics, 16(8), 3386-3398 (2019-06-27)
Clinical trials involving p53 gene therapy for ovarian cancer failed due to the dominant negative inhibition of wild-type p53 and multiple genetic aberrations in ovarian cancer. To overcome this problem, we have designed a more potent chimeric gene fusion, called
Kyle Trudeau et al.
Investigative ophthalmology & visual science, 52(12), 8657-8664 (2011-10-08)
Mitochondrial dysfunction is known to play a role in retinal vascular cell loss, a prominent lesion of diabetic retinopathy. High glucose (HG) has been reported to induce mitochondrial fragmentation and dysfunction in retinal endothelial cells, contributing to apoptosis. In this

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service