Skip to Content
Merck
All Photos(1)

Key Documents

I1886

Sigma-Aldrich

Anti-Human IgG (whole molecule) antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Human IgG, Goat Anti-Human IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect ELISA: 1:10,000
quantitative precipitin assay: 3.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders . Anti-human IgG (whole molecule) antibody is specific for human IgG when tested against normal human serum and IgG by immunoelectrophoresis. The antibody also reacts with light chains. Immunoglobulin G (IgG) is part of the immunoglobulin family and is a widely expressed serum antibody. It consists of a γ heavy chain in the constant (C) region. The primary structure of this antibody contains disulfide bonds involved in linking the two heavy chains, linking the heavy and light chains and also resides inside the chains.

Immunogen

Human IgG

Application

Anti-Human IgG (whole molecule) antibody is suitable for use in quantitative precipitin assay (3.0 mg/mL).
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuerhongjiang Tuxun et al.
Scientific reports, 8(1), 4417-4417 (2018-03-15)
Fluorodeoxyglucose (FDG) uptake by alveolar echinococcosis (AE) liver lesions is a signal of their metabolic activity and of disease progression. In order to find a surrogate marker for this status, we investigated whether parameters of the peripheral and/or periparasitic immune
Laura E M Dunn et al.
Journal of virology, 97(2), e0189422-e0189422 (2023-02-07)
The ability of Epstein-Barr virus (EBV) to switch between latent and lytic infection is key to its long-term persistence, yet the molecular mechanisms behind this switch remain unclear. To investigate transcriptional events during the latent-to-lytic switch, we utilized Precision nuclear
Lorenzo Russo et al.
Nanoscale, 11(22), 10819-10827 (2019-05-29)
Myxovirus protein A (MxA) is a biomarker that can be used to distinguish between viral and bacterial infections. While MxA lateral flow assays (LFAs) have been successfully used for viral vs. bacterial differential diagnosis for children, the clinically relevant level
Yakov A Lomakin et al.
Communications biology, 7(1), 842-842 (2024-07-11)
Identifying high-affinity antibodies in human serum is challenging due to extremely low number of circulating B cells specific to the desired antigens. Delays caused by a lack of information on the immunogenic proteins of viral origin hamper the development of
Nur Alam et al.
Cellulose (London, England), 1-11 (2021-07-27)
Lateral flow assay (LFA) is an important point-of-care (POC) test platform due to the associated portability, on-site testing, and low cost for diagnosis of pathogen infections and disease biomarkers. However, compared to high-end analyzers in hospitals, LFA devices, in particular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service