Skip to Content
Merck
All Photos(1)

Documents

I0759

Sigma-Aldrich

Anti-Human IgM (μ-chain specific) antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti Human IgM Antibody - Anti-Human IgM (μ-chain specific) antibody produced in goat, Anti Human Igm, Anti Human Igm Antibody, Anti-Human Igm

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: suitable
quantitative precipitin assay: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Human serum-derived IgM may be used as a reference antigen, standard, blocking agent, or coating protein in a variety of immunoassays including ELISA, dot immunobinding, Western immunoblotting, immunodiffusion, and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solid phase immunoadsorbents.
Immunoglobulin M (IgM) antibodies appear early in the course of infections. IgM antibodies are responsible for agglutination of red blood cells in mis-matched blood transfusions. The level of IgM may vary with the status of disease or infection.
IgM antibodies are present as pentamers in the serum and are produced in response to antigens
Anti-Human IgM (μ-chain specific) antibody is specific for human IgM with mu chain. The antibody preparation is specific for human IgM when tested against purified human IgA, IgG, IgM, Bence Jones κ and Bence Jones λ myeloma proteins.

Specificity

Goat polyclonal Anti-Human IgM (μ-chain specific) antibody reacts with the m-chain of human IgM. It is specific for human IgM when tested against purified human IgA, IgG, IgM, Bence Jones kappa and Bence Jones lambda myeloma proteins.

Immunogen

Purified human IgM as immunogen

Application

Anti-Human IgM (μ-chain specific) antibody is suitable for Ouchterlony double diffusion and quantitative precipitin assay. It was used in B-cell receptor cross-linking studies, AuBioDOT assays, ELISA, protein purification and gel blot analysis.
Goat polyclonal Anti-Human IgM (μ-chain specific) antibody may be used to identify and quantitate the level of human IgM in vitro and in human serum or other biological fluids by immunochemical or immunohistological techniques.

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Vázquez et al.
Clinical and diagnostic laboratory immunology, 10(6), 1074-1077 (2003-11-11)
The Dengue IgM Capture ELISA (MAC-ELISA) is the immunoenzymatic system recommended by the Pan American Health Organization and the World Health Organization for the serological diagnosis of dengue virus infection due to its high sensitivity, ease of performance, and use
Zhen Lin et al.
Journal of virology, 90(7), 3469-3479 (2016-01-15)
In the oral epithelium, peripheral stores of Epstein-Barr virus (EBV) are transmitted from infiltrating B cells to epithelial cells. Once the virus is transmitted to epithelial cells, the highly permissive nature of this cell type for lytic replication allows virus
Harini Natarajan et al.
BMC immunology, 23(1), 7-7 (2022-02-18)
While antibodies can provide significant protection from SARS-CoV-2 infection and disease sequelae, the specific attributes of the humoral response that contribute to immunity are incompletely defined. We employ machine learning to relate characteristics of the polyclonal antibody response raised by
Qinyan Yin et al.
Virology, 327(1), 134-143 (2004-08-26)
Transcription of the immediate early gene BZLF1 is mediated initially through the activation of cellular transcription factors. Reporter-based studies have provided evidence that following this initial activation, the BZLF1 gene product Zta may be involved in an autoactivation loop through
John G Kosowicz et al.
Journal of virology, 91(16) (2017-06-02)
Epstein-Barr virus (EBV) is a ubiquitous human gammaherpesvirus that establishes a latency reservoir in B cells. In this work, we show that ibrutinib, idelalisib, and dasatinib, drugs that block B cell receptor (BCR) signaling and are used in the treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service