Skip to Content
Merck
All Photos(1)

Documents

I1020

Sigma-Aldrich

Interleukin-4 from mouse

IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

mIL-4, Interleukin-4 human

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

0.1-2.0 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

~13.5 kDa

packaging

pkg of 5 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Il4(16189)

Application

Interleukin-4 from mouse has been used:
  • as a component of cell culture media for class switch recombination assay
  • to study its therapeutic benefits for mucosal healing
  • as a pioglitazone or new peroxisome proliferator-activated receptor agonist (GQ-11)

Biochem/physiol Actions

Interleukin-4 (IL-4) is a lymphokine with profound effects on the growth and differentiation of immunologically competent cells. IL-4 is also known as B cell stimulatory factor-1 (BSF-1), T cell growth factor-2 (TCGF-2) and mast-cell growth factor-2 (MCGF-2). Inhibits VEGF-induced and FGF-Basic-induced angiogenesis. IL-4 is a complex glycoprotein released by a subset of activated T cells. Treatment of IL-4 with specific glycosidases yields an active polypeptide. Human and mouse IL-4 share 50% amino acid sequence homology, but their biological actions are species-specific.

Physical form

Lyophilized, 0.2 μm filtered, with no additives.

Analysis Note

The proliferative activity is tested in culture using mouse HT-2 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin 4 induces rapid mucin transport, increases mucus thickness and quality and decreases colitis and Citrobacter rodentium in contact with epithelial cells
Sharba S, et al.
Virulence, 10(1), 97-117 (2019)
VX-984 is a selective inhibitor of non-homologous end joining, with possible preferential activity in transformed cells
Khan AJ, et al.
Oncotarget, 9(40), 25833-25833 (2018)
New Peroxisome Proliferator-Activated Receptor Agonist (GQ-11) Improves Wound Healing in Diabetic Mice
Silva JC, et al.
Advances in Wound Care (2019)
R Fernandez-Botran et al.
The Journal of experimental medicine, 164(2), 580-593 (1986-08-01)
T cell-derived supernatants (SN) that contain B cell-stimulatory factor 1 (BSF-1) and lack IL-2 promote the growth of the IL-2-dependent T cell line, HT-2, as well as three other clones or lines of T cells that can provide help to
Saeko Nakajima et al.
The Journal of investigative dermatology, 134(8), 2122-2130 (2014-02-01)
Atopic dermatitis (AD) is generally regarded as a type 2 helper T (Th2)-mediated inflammatory skin disease. Although the number of IL-17A-producing cells is increased in the peripheral blood and in acute skin lesion of AD patients, the role of IL-17A

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service