Merck
  • IL-17A as an inducer for Th2 immune responses in murine atopic dermatitis models.
IL-17A as an inducer for Th2 immune responses in murine atopic dermatitis models.
The Journal of investigative dermatology (2014-02-01)
Saeko Nakajima, Akihiko Kitoh, Gyohei Egawa, Yohei Natsuaki, Satoshi Nakamizo, Catharina Sagita Moniaga, Atsushi Otsuka, Tetsuya Honda, Sho Hanakawa, Wataru Amano, Yoichiro Iwakura, Susumu Nakae, Masato Kubo, Yoshiki Miyachi, Kenji Kabashima
ABSTRACT
Atopic dermatitis (AD) is generally regarded as a type 2 helper T (Th2)-mediated inflammatory skin disease. Although the number of IL-17A-producing cells is increased in the peripheral blood and in acute skin lesion of AD patients, the role of IL-17A in the pathogenesis of AD remains unclear. To clarify this issue, we used murine AD models in an IL-17A-deficient condition. In a repeated hapten application-induced AD model, skin inflammation, IL-4 production in the draining lymph nodes (LNs), and hapten-specific IgG1 and IgE induction were suppressed in IL-17A-deficient mice. Vγ4(+) γδ T cells in the skin-draining LNs and Vγ5(-) dermal γδ T cells in the skin were the major sources of IL-17A. Consistently, in flaky-tail (Flg(ft/ft) ma/ma) mice, spontaneous development of AD-like dermatitis and IgE induction were attenuated by IL-17A deficiency. Moreover, Th2 differentiation from naive T cells was promoted in vitro by the addition of IL-17A. Taken together, our results suggest that IL-17A mediates Th2-type immune responses and that IL-17A signal may be a therapeutic target of AD.
Sigma-Aldrich
Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture