Skip to Content
Merck
All Photos(6)

Key Documents

HPA039410

Sigma-Aldrich

Anti-UBE3A antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ANCR, Anti-AS, Anti-E6-AP, Anti-EPVE6AP, Anti-FLJ26981, Anti-HPVE6A, Anti-Ubiquitin protein ligase E3A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HLIERYYHQLTEGCGNEACTNEFCASCPTFLRMDNNAAAIKALELYKINAKLCDPHPSKKGASSAYLENSKGAPNNSCSEIK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE3A(7337)

General description

The ubiquitin protein ligase E3A (UBE3A) gene is mapped to human chromosome 15q11-q13. UBE3A codes for E6-AP ubiquitin-protein ligase, which belongs to E3 ubiquitin ligase protein family. The members of this family consist of a homologous to E6AP C-terminus (HECT) domain involved in the transfer of ubiquitin from E2 to the substrate.

Immunogen

ubiquitin protein ligase E3A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The ubiquitin protein ligase E3A (UBE3A) functions as an E3 ligase in the ubiquitin-proteasome pathway and acts as a transcriptional co-activator of steroid hormone receptors such as progesterone and estrogen. During papillomavirus infection, ubiquitin protein ligase E3A (UBE3A), along with E6 viral protein, plays a vital role in degradation of the cell cycle regulatory protein p53. Anomalistic degradation of p53 can cause cervical cancer. Inactivation or deletion of UBE3A gene leads to the neurodevelopmental disorder Angelman syndrome (AS), while overexpression of maternal UBE3A gene is associated with autism pathogenesis. Therefore, UBE3A level has to be properly controlled for normal brain development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81436

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

De novo truncating mutations in E6-AP ubiquitin-protein ligase gene (UBE3A) in Angelman syndrome.
Matsuura T
Nature Genetics, 15, 74-77 (1997)
Epigenetic regulation of UBE3A and roles in human neurodevelopmental disorders.
LaSalle JM
Epigenomics, 7, 1213-1228 (2015)
An Autism-Linked Mutation Disables Phosphorylation Control of UBE3A
Yi JJ
Cell, 162, 795-807 (2015)
Catalytically Important Residues of E6AP Ubiquitin Ligase Identified Using Acid-Cleavable Photo-Cross-Linkers.
Krist DT and Statsyuk AV
Biochemistry, 54, 4411-4414 (2015)
The Angelman syndrome ubiquitin ligase localizes to the synapse and nucleus, and maternal deficiency results in abnormal dendritic spine morphology.
Dindot SV
Human Molecular Genetics, 17, 111-118 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service