Skip to Content
Merck
All Photos(6)

Key Documents

HPA001562

Sigma-Aldrich

Anti-ATF3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

ATF3 Antibody - Anti-ATF3 antibody produced in rabbit, Atf3 Antibody, Anti-Activating transcription factor 3 antibody produced in rabbit, Anti-Cyclic AMP-dependent transcription factor ATF-3 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MMLQHPGQVSASEVSASAIVPCLSPPGSLVFEDFANLTPFVKEELRFAIQNKHLCHRMSSALESVTVSDRPLGVSITKAEVAPEEDERKKRRRERNKIAAAKCRNKKKEKTEC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATF3(467)

General description

Activating transcription factor 3 (ATF3) gene is mapped to human chromosome 1q32.3. The encoded protein belongs to the cAMP-response element binding (CREB) protein family.
Rabbit polyclonal anti-ATF3 antibody reacts with human activating transcription factor 3.

Immunogen

Cyclic AMP-dependent transcription factor ATF-3 recombinant protein epitope signature tag (PrEST)

Application

Anti-ATF3 antibody produced in rabbit has been used in:
  • western blotting
  • immunofluorescence
  • immunohistochemical staining
  • chromatin immunoprecipitation (ChIP)

Biochem/physiol Actions

Activating transcription factor 3 (ATF3), a cyclic AMP-dependent transcription factor stimulates transcription by sequestering inhibitory cofactors. ATF3 responds to stress signals and is involved in the regulation of immune and metabolic homeostasis. It helps to prevent chronic inflammation and starvation responses. ATF3 expression is induced in response to eye injury. It serves as a marker for neuronal injury.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77475

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A potential dichotomous role of ATF3, an adaptive-response gene, in cancer development
Yin X, et al.
Oncogene, 27(15), 2118-2118 (2008)
Marta Bueno et al.
Aging cell, 17(2) (2018-01-25)
PINK1 (PTEN-induced putative kinase 1) is a key regulator of mitochondrial homeostasis that is relatively depleted in aging lungs and in lung epithelial cells from patients with idiopathic pulmonary fibrosis (IPF), a disease linked with aging. Impaired PINK1 expression and
Activating transcription factor 3 promotes embryo attachment via up-regulation of leukemia inhibitory factor in vitro
Cheng X, et al.
Reproductive Biology and Endocrinology, 15(1), 42-42 (2017)
Nihal Kaplan et al.
Investigative ophthalmology & visual science, 60(10), 3570-3583 (2019-08-17)
Single-cell RNA-sequencing (scRNA-seq) was used to interrogate the relatively rare stem (SC) and early transit amplifying (TA) cell populations in limbal/corneal epithelia from wild-type and autophagy-compromised mice. We conducted scRNA-seq on ocular anterior segmental tissue from wild-type and beclin 1-deficient
Margaret B Allison et al.
Diabetes, 67(6), 1093-1104 (2018-03-15)
Leptin acts via its receptor (LepRb) to modulate gene expression in hypothalamic LepRb-expressing neurons, thereby controlling energy balance and glucose homeostasis. Despite the importance of the control of gene expression in hypothalamic LepRb neurons for leptin action, the transcriptional targets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service