Skip to Content
Merck
All Photos(2)

Key Documents

HPA021520

Sigma-Aldrich

Anti-SVEP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-CCP module-containing protein 22, Anti-Polydom, Anti-SEL-OB, Anti-Selectin-like osteoblast-derived protein, Anti-Serologically defined breast cancer antigen NY-BR-38, Anti-Sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

GDFICTPDNTGVNCTLTCLEGYDFTEGSTDKYYCAYEDGVWKPTYTTEWPDCAKKRFANHGFKSFEMFYKAARCDDTDLMKKFSEAFETTLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SVEP1(79987)

General description

SVEP1 (Sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1) also called as polydom or SEL-OB (selectin-like osteoblast-derived protein) is expressed primarily in osteogenic cells. The gene coding for protein is localized to human chromosome 9q32.
SVEP1 protein consists of multiple domains such as vWA (von Willebrand factor type A), EGF (epidermal growth factor), PTX (pentraxin), and HYR (hyalin repeat) domains, and complement control protein (CCP) modules which are also called sushi domains.

Immunogen

Sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SVEP1 (Sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1) plays a vital role in cell adhesion process and immune response mediated by complement system. This secretory protein serves as a marker for pre-fated satellite cells of skeletal mucle. SVEP1 protein acts as a ligand for integrin α9β1 receptor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75350

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabi Shefer et al.
Stem cell reviews, 6(1), 42-49 (2010-01-07)
In this study we explored the expression pattern of SVEP1, a novel cell adhesion molecule (CAM), in bona fide satellite cells and their immediate progeny. We show that SVEP1 is expressed in activated satellite cells prior to their determination to
I Shur et al.
Journal of cellular physiology, 206(2), 420-427 (2005-10-06)
We describe a novel human gene, named SEL-OB/SVEP1, expressed by skeletal tissues in vivo and by cultured osteogenic cells. The mRNA expression was analyzed on frozen tissues retrieved by laser-capture microscope dissection (LCM) and was detected in osteogenic tissues (periosteum
Ryoko Sato-Nishiuchi et al.
The Journal of biological chemistry, 287(30), 25615-25630 (2012-06-02)
A variety of proteins, including tenascin-C and osteopontin, have been identified as ligands for integrin α9β1. However, their affinities for integrin α9β1 are apparently much lower than those of other integrins (e.g. α3β1, α5β1, and α8β1) for their specific ligands
D Gilgès et al.
The Biochemical journal, 352 Pt 1, 49-59 (2000-11-04)
To identify extracellular proteins with epidermal growth factor (EGF) domains that are potentially involved in the control of haemopoiesis, we performed degenerate reverse-transcriptase-mediated PCR on the murine bone-marrow stromal cell line MS-5 and isolated a new partial cDNA encoding EGF-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service