Skip to Content
Merck
All Photos(1)

Key Documents

MABT26

Sigma-Aldrich

Anti-E-cadherin Antibody, clone DECMA-1

clone Decma-1, from rat

Synonym(s):

Anti-Anti-Arc-1, Anti-Anti-BCDS1, Anti-Anti-CD324, Anti-Anti-CDHE, Anti-Anti-ECAD, Anti-Anti-LCAM, Anti-Anti-UVO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Decma-1, monoclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)
mouse ... Cdh1(12550)

General description

Cadherins are a family of transmembrane glycoproteins involved in Ca2+-dependent cell to cell interactions. The members of the family are differentially expressed in various tissues and function in the maintenance of tissue integrity, morphogenesis, and migration. Cadherins are divided into type I and type II subgroups. Type I cadherins include epithelial cadherin (E-cadherin), neural cadherin (N-cadherin), placental cadherin (P-cadherin) and retinal cadherin (R-cadherin). Type II include kidney cadherin (K-cadherin) and osteoblast cadherin (OB-cadherin). One of the best characterized cadherins is E-cadherin (epithelial cadherin or cadherin-1). It consists of calcium-binding extracellular domains responsible for interactions with other E-cadherin molecules on neighboring cells and interaction with integrin alphaEbeta7. The cytoplasmic domain of E-cadherin is linked to the actin cytoskeleton through cytoplasmic catenin proteins, thus establishing a complex localized to adherens junctions. In carcinomas, E-cadherin is frequently downregulated, consistent with its function of an invasion suppressor in normal epithelia.

Immunogen

Mouse embryonal carcinoma cell line PCC4

Application

Immunohistochemistry Analysis: 1:300 dilution of this antibody has been shown to detect E-cadherin in mouse GI tract tissue.

Inhibition: A previous lot was used by an independent laboratory in inhibion studies, blocking E-cadherin associated cell adhesion. (Vestweber, 1985)

Flow Cytometry Analysis: A previous lot was used by an independent laboratory in flow cytometry applications. (Tang, 1994)
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-E-cadherin Antibody, clone DECMA-1 is validated for use in WB, IH, IC, IP, FC, FUNC for the detection of E-cadherin.

Quality

Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected E-Cadherin in 10 µg of A431 cell lysate.

Target description

~ 106 kDa observed MW.

Physical form

Format: Purified
Protein G
Purified rat monoclonal IgG1κ in buffer containing PBS without preservative.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detian Yuan et al.
Cancer cell, 31(6), 771-789 (2017-06-14)
Intrahepatic cholangiocarcinoma (ICC) is a highly malignant, heterogeneous cancer with poor treatment options. We found that mitochondrial dysfunction and oxidative stress trigger a niche favoring cholangiocellular overgrowth and tumorigenesis. Liver damage, reactive oxygen species (ROS) and paracrine tumor necrosis factor
S Pece et al.
The Journal of biological chemistry, 275(52), 41227-41233 (2000-09-02)
E-cadherins are well characterized cell surface molecules expressed in epithelial cells, which play a major role in cell adhesion through the establishment of calcium-dependent homophilic interactions at sites of cell-cell contacts. They are also integral components of morphogenetic programs controlling
Nicholas F Dybdal-Hargreaves et al.
Oncotarget, 10(45), 4719-4720 (2019-08-07)
[This corrects the article DOI: 10.18632/oncotarget.23798.].
Philippe Pujuguet et al.
Molecular biology of the cell, 14(5), 2181-2191 (2003-06-13)
Ezrin, a membrane cytoskeleton linker, is involved in cellular functions, including epithelial cell morphogenesis and adhesion. A mutant form of ezrin, ezrin T567D, maintains the protein in an open conformation, which when expressed in Madin-Darby canine kidney cells causes extensive
Zhibo Gai et al.
Journal of the American Society of Nephrology : JASN, 21(9), 1468-1476 (2010-05-29)
Mutations in TRPS1 cause tricho-rhino-pharyngeal syndrome (TRPS). Trps1 is essential for nephron development, acting downstream of Bmp7. Because Bmp7 counteracts epithelial-to-mesenchymal transition (EMT) and reverses chronic renal injury, we examined the function of Trps1 in renal fibrosis. Immunohistochemistry revealed Trps1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service