Skip to Content
Merck
All Photos(2)

Key Documents

HPA018007

Sigma-Aldrich

Anti-ITSN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Intersectin-1, Anti-SH3 domain-containing protein 1A, Anti-SH3P17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ITVLEQQDMWWFGEVQGQKGWFPKSYVKLISGPIRKSTSMDSGSSESPASLKRVASPAAKPVVSGEEFIAMYTYESSEQGDLTFQQGDVILVTKKDGDWW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITSN1(6453)

General description

Intersectin 1 (ITSN1) belongs to the intersectin (ITSN1) family. The scaffolding protein has two Eps15 homology (EH) domains, a coiled-coil domain and five Src homology 3 (SH3) domains. The gene encoding ITSN1 is localized on human chromosome 21.

Immunogen

Intersectin-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Intersectin 1 (ITSN1) is involved in clathrin-mediated endocytosis wherein it aids in clustering of membrane-sculpting proteins in the early stages. During vaccinia virus infection, it activates the cell division control protein 42 homolog (Cdc42). It also has a role in phagocytosis mediated by the Fc-γ receptor and regulates many signal transduction pathways like those involving phosphatidylinositol 3-kinase. ITSN1 has been shown to be expressed in neuroblastoma tumors and tumor cell lines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73912

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Novokhatska et al.
PloS one, 8(7), e70546-e70546 (2013-08-13)
Scaffolding proteins of the intersectin (ITSN) family, ITSN1 and ITSN2, are crucial for the initiation stage of clathrin-mediated endocytosis. These proteins are closely related but have implications in distinct pathologies. To determine how these proteins could be separated in certain
Alba Zuidema et al.
Journal of cell science, 131(21) (2018-10-12)
The family of integrin transmembrane receptors is essential for the normal function of multicellular organisms by facilitating cell-extracellular matrix adhesion. The vitronectin-binding integrin αVβ5 localizes to focal adhesions (FAs) as well as poorly characterized flat clathrin lattices (FCLs). Here, we
A Russo et al.
Oncogene, 31(46), 4828-4834 (2012-01-24)
Intersectin 1 (ITSN1) is a scaffold protein that regulates diverse cellular pathways, including endocytosis and several signal transduction pathways, including phosphatidylinositol 3-kinase, Class IIβ (PI3K-C2β). ITSN1's transforming potential in vitro suggests that this scaffold protein may be involved in human
Michael P Hunter et al.
Neuroreport, 22(15), 767-772 (2011-08-31)
Intersectin 1 (ITSN1) is a human chromosome 21 (HSA21) gene product encoding a multidomain scaffold protein that functions in endocytosis, signal transduction, and is implicated in Down's syndrome, Alzheimer's Disease, and potentially other neurodegenerative diseases through activation of c-Jun N-terminal
Xinxin Wang et al.
eLife, 9 (2020-05-01)
Clathrin-mediated endocytosis (CME) in mammalian cells is driven by resilient machinery that includes >70 endocytic accessory proteins (EAP). Accordingly, perturbation of individual EAPs often results in minor effects on biochemical measurements of CME, thus providing inconclusive/misleading information regarding EAP function.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service