Skip to Content
Merck
All Photos(1)

Key Documents

HPA003892

Sigma-Aldrich

Anti-RARRES1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LXNL, Anti-TIG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VVFSTERYNPESLLQEGEGRLGKCSARVFFKNQKPRPTINVTCTRLIEKKKRQQEDYLLYKQMKQLKNPLEIVSIPDNHGHIDPSLRLIW

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RARRES1(5918)

Immunogen

Retinoic acid receptor responder protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Retinoic acid receptor responder protein 1 is a protein encoded by the RARRES1 gene in humans. It is a functionally undefined membrane protein that confers shorter survival in patients with Inflammatory breast cancer (IBC). Depletion of this protein inhibits tumor growth of IBC cells in vivo and decreases IBC cell proliferation, migration and invasion in vitro. This gene expression by promoter hypermethylation may play an important role in HCC (hepatocellular carcinoma). It is a tumor suppressor gene and is frequently hypermethylated in several cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86679

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi-Hua Chen et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(2), 967-971 (2013-09-06)
Recently, it has been reported that tazarotene-induced gene 1 (TIG1) methylation was frequently detected in a variety of human cancers. However, the relationship between the TIG1 methylation and the characteristics of hepatocellular carcinoma (HCC) remains unknown. The aim of present
Xiaoping Wang et al.
Cancer research, 73(21), 6516-6525 (2013-09-10)
Inflammatory breast cancer (IBC) is the most lethal form of breast cancer, but the basis for its aggressive properties are not fully understood. In this study, we report that high tumoral expression of TIG1 (RARRES1), a functionally undefined membrane protein
Arpita Roy et al.
PloS one, 12(7), e0180344-e0180344 (2017-07-06)
Prostate cancer (PCa) poses a major health concern in men worldwide. Retinoic Acid Receptor Responder (RARRES1)/ Tazarotene-induced gene-1 (TIG-1) is a putative tumor suppressor gene that exerts its tumor suppressor function via unknown mechanisms. Epigenetic silencing of RARRES1 leads to
Michael Kloth et al.
International journal of cancer, 131(6), E897-E904 (2012-05-11)
Epigenetic aberrations are frequent in prostate cancer and could be useful for detection and prognostication. However, the underlying mechanisms and the sequence of these changes remain to be fully elucidated. The tumor suppressor gene RARRES1 (TIG1) is frequently hypermethylated in
Lehel Peterfi et al.
British journal of cancer, 122(12), 1818-1824 (2020-04-21)
Approximately 15% of clinically localised conventional renal cell carcinoma (RCC) will develop metastasis within 5 years of follow-up. The aim of this study was to identify biomarkers predicting the postoperative tumour relapse. Tissue microarrays of conventional RCC from a cohort

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service