Skip to Content
Merck
All Photos(6)

Key Documents

HPA002823

Sigma-Aldrich

Anti-TM4SF1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-M3S1 antibody produced in rabbit, Anti-Membrane component surface marker 1 antibody produced in rabbit, Anti-Transmembrane 4 L6 family member 1 antibody produced in rabbit, Anti-Tumor-associated antigen L6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

GPLCLDSLGQWNYTFASTEGQYLLDTSTWSECTEPKHIVEWNVSLFSI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TM4SF1(4071)

Related Categories

Immunogen

Transmembrane 4 L6 family member 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-TM4SF1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Transmembrane 4 L6 family member 1 is a protein encoded by the TM4SF1 gene in humans. It is a cell surface antigen, which is highly expressed in lung, breast, colon and ovarian carcinomas. It has a novel function in Androgen Receptor (AR) signaling. Its mRNA expression is higher in prostate cancer tissues as compared to benign prostatic hyperplasia (BPH). It is found to contribute to prostate cancer cell metastasis. The gene serves as a molecular organizer that interacts with membrane and cytoskeleton-associated proteins. It uniquely initiates the formation of nanopodia and facilitates cell polarization and migration. It also regulates tumour cell motility and invasiveness. It is abundant on the plasma membrane and on intracellular vesicles.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85203

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathalie Allioli et al.
The Prostate, 71(11), 1239-1250 (2011-06-10)
The Androgen Receptor (AR) plays a key role in controlling prostate gland homeostasis and contributes to prostate carcinogenesis. The identification of its target genes should provide new candidates that may be implicated in cancer initiation and progression. Transcriptomic experiments and
J S Marken et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(8), 3503-3507 (1992-04-15)
The L6 cell surface antigen, which is highly expressed on lung, breast, colon, and ovarian carcinomas, has attracted attention as a therapeutic target for murine monoclonal antibodies and their humanized counterparts. Its molecular nature has, however, remained elusive. Here we
Andrew Zukauskas et al.
Angiogenesis, 14(3), 345-354 (2011-06-01)
Transmembrane-4-L-six-family-1 (TM4SF1) is a tetraspanin-like membrane protein that is highly and selectively expressed by cultured endothelial cells (EC) and, in vivo, by EC lining angiogenic tumor blood vessels. TM4SF1 is necessary for the formation of unusually long (up to a
Tamara Lekishvili et al.
Journal of cell science, 121(Pt 5), 685-694 (2008-02-14)
Tumour-associated antigen L6 (L6-Ag, also known as TM4SF1) regulates tumour cell motility and invasiveness. We found that L6-Ag is abundant on the plasma membrane and on intracellular vesicles, on which it is co-localised with the markers for late endosomal/lysosomal compartments
Biao Zheng et al.
International journal of oncology, 47(2), 490-498 (2015-06-03)
The cell surface protein Transmembrane 4 L6 family member 1 (TM4SF1) has been detected in various tumors, and its expression on tumor cells is implicated in cancer cell metastasis and patient prognosis. The role of TM4SF1 in malignant tumors remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service