Skip to Content
Merck
All Photos(7)

Documents

HPA002689

Sigma-Aldrich

Anti-WASHC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FAM39E, Anti-FLJ00038, Anti-WASH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GAPREVVDPSGGWATLLESIRQAGGIGKAKLRSMKERKLEKKKQKEQEQVRATSQGGHLMSDLFNKLVMRHKGISGKGPGAGEGPGGAFARVSDSIPPLPPPQQPQVDEDEDDWES

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

WASH complex subunit 1 (WASHC1) is located on human chromosome 9p24.3. It belongs to Wiskott-Aldrich Syndrome Protein and SCAR Homolog (WASH) family. WASHC1 protein has an actin binding domain.

Immunogen

WASH complex subunit 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-WASHC1 antibody produced in rabbit has been used:
  • in the detection of WASH complex in Dictyostelium cells by immunofluorescence imaging.
  • in the detection of WASH proteins in HeLa cells using confocal microscopy.
  • in western blotting for the detection of WASH1 complexes in human leukemic Jurkat T cells.

Biochem/physiol Actions

WASH exists as multiple complex protein in HeLa and bovine cells. WASH proteins coordinate the migration of endosomal α5β1 integrin. WASHC1 protein regulates actin cytoskeleton organisation. WASH1 protein is regarded as actin-nucleating promoting factor. WASH protein complex interacts with actin related protein, Arp2/3 and mediates actin networking.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74318

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human subtelomeric WASH genes encode a new subclass of the WASP family
Linardopoulou EV, et al.
PLoS Genetics, 3(12), e237-e237 (2007)
Functional characterization of Wiskott Aldrich Syndrome protein and scar homolog (WASH), a bi-modular nucleation promoting factor (NPF) able to interact with biogenesis of lysosome related organelle subunit 2 (BLOS-2) and gamma-tubulin
Monfregola J, et al.
The Journal of Biological Chemistry, jbc-M109 (2010)
The Arp2/3 activator WASH regulates alpha5beta1-integrin-mediated invasive migration
Zech T, et al.
Journal of Cell Science, 124(22), 3753-3759 (2011)
WASH drives early recycling from macropinosomes and phagosomes to maintain surface phagocytic receptors
Buckley CM, et al.
Proceedings of the National Academy of Sciences of the USA, 113(40), E5906-E5915 (2016)
The Arp2/3 activator WASH controls the fission of endosomes through a large multiprotein complex
Derivery E, et al.
Developmental Cell, 17(5), 712-723 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service