Skip to Content
Merck
All Photos(2)

Documents

ABT79

Sigma-Aldrich

Anti-WASH complex subunit FAM21C Antibody

from rabbit

Synonym(s):

family with sequence similarity 21, member C, vaccinia virus penetration factor, WASH complex subunit FAM21C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WASHC4(23325)

General description

FAM21C, also known as WASH complex subunit FAM21C, is a part of the a 500 kDa core complex that allows the Wiskott-Aldrich syndrome protein (WASH) function. FAM21C can be found in the membranes of early endosomes. FAM21C cooperates with CAPZ and hinders its actin-capping activity, links WASH to endosomes, and is required for WASH-dependent retromer-mediated sorting.

Immunogen

Recombinant protein corresponding to human WASH complex subunit FAM21C.

Application

Anti-WASH complex subunit FAM21C Antibody detects level of WASH complex subunit FAM21C & has been published & validated for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilutioin from a previous lot detected WASH complex subunit FAM21C in NIH/3T3, A431, and HeLa cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeletal Signaling

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 5 µg/mL of this antibody detected WASH complex subunit FAM21C in 10 µg of HEK293 cell lysate.

Target description

~145 kDa observed
Non-specific bands may be observed >200 kDa and at ~10 kDa

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew N J Seaman et al.
Journal of cell science, 131(12) (2018-05-20)
The retromer complex is a vital component of the endosomal protein sorting machinery necessary for sorting into both the endosome-to-Golgi retrieval pathway and also the endosome-to-cell-surface recycling pathway. Retromer mediates cargo selection through a trimeric complex comprising VPS35, VPS29 and
Rakesh Kulkarni et al.
Life science alliance, 6(4) (2023-01-31)
FAM21 (family with sequence similarity 21) is a component of the Wiskott-Aldrich syndrome protein and SCAR homologue (WASH) protein complex that mediates actin polymerization at endosomal membranes to facilitate sorting of cargo-containing vesicles out of endosomes. To study the function
Yi Cui et al.
Traffic (Copenhagen, Denmark), 22(4), 123-136 (2020-12-22)
Retromer core complex is an endosomal scaffold that plays a critical role in orchestrating protein trafficking within the endosomal system. Here we characterized the effect of the Parkinson's disease-linked Vps35 D620N in the endo-lysosomal system using Vps35 D620N rescue cell
Helen Chen et al.
JCI insight, 5(17) (2020-09-04)
Prader-Willi syndrome (PWS) is a developmental disorder caused by loss of maternally imprinted genes on 15q11-q13, including melanoma antigen gene family member L2 (MAGEL2). The clinical phenotypes of PWS suggest impaired hypothalamic neuroendocrine function; however, the exact cellular defects are
Jenai Quan et al.
mBio, 15(8), e0144524-e0144524 (2024-07-02)
Neurotropic alphaherpesviruses, including herpes simplex virus type 1 and pseudorabies virus, establish a lifelong presence within the peripheral nervous system of their mammalian hosts. Upon entering cells, two conserved tegument proteins, pUL36 and pUL37, traffic DNA-containing capsids to nuclei. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service