Skip to Content
Merck
All Photos(10)

Key Documents

HPA000834

Sigma-Aldrich

Anti-G6PD antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-G-6-PDH antibody produced in rabbit, Anti-G6PDH antibody produced in rabbit, Anti-Glucose-6-phosphate dehydrogenase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

FQGDAFHQSDTHIFIIMGASGDLAKKKIYPTIWWLFRDGLLPENTFIVGYARSRLTVADIRKQSEPFFKATPEEKLKLEDFFARNSYVAGQYDDAASYQRLNSHMNALH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G6PD(2539)

Looking for similar products? Visit Product Comparison Guide

General description

G6PD gene is mapped to human chromosome Xq28, and spans 16.2kb. Its deficiency is highly hetereogenous, with around 190 variants being reported.
Glucose-6-phosphate dehydrogenase (G6PD), a cytoplasmic enzyme, is expressed in all the tissues.

Immunogen

Glucose-6-phosphate dehydrogenase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-G6PD antibody is suitable for:
  • chromatin immunoprecipitation (CHIP)
  • immunofluorescence
  • western blotting

Anti-G6PD antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link.To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Biochem/physiol Actions

Glucose-6-Phosphate Dehydrogenase (G6PD) deficiency is recessively inherited predominantly in males. Its activity shows statistical significant correlation with total bilirubin blood levels. G6PD suppression may help in the treatment strategies for inhibiting tumor progression and acts as a novel anti-hepatocellular carcinoma (HCC) target. Its deficiency is a common inherited enzyme defect (a disorder associated with acute or chronic hemolytic anemia). G6PD enzyme is responsible for the oxidation of glucose-6-phosphate, and thus, confers protection against oxidative damage in erythrocytes.
Glucose-6-phosphate dehydrogenase (G6PD), the rate-limiting enzyme of the pentose phosphate pathway (PPP), plays a major role in cells to resist oxidative stress. In humans, G6PD deficiency is the prevalent enzymopathy, which causes hemolytic anemia and newborn jaundice. G6PD is involved in the accumulation of reactive oxygen species (ROS). It is also involved in the progression of various cancer, including renal cell carcinoma (RCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74032

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natsuda Jamornthanyawat et al.
PloS one, 9(2), e88605-e88605 (2014-03-04)
Glucose-6-phosphate dehydrogenase (G6PD) deficiency is a common inherited enzyme defect and an important problem in areas with Plasmodium vivax infection because of the risk of haemolysis following administration of primaquine to treat the liver forms of the parasite. We undertook
Xue-Jiang Gu et al.
Acta diabetologica, 50(1), 89-92 (2010-11-17)
A 59-year-old Chinese male patient was admitted at diagnosis of type 1 diabetes with ketoacidosis. During the normalization of blood glucose with insulin, the patient developed acute hemolysis. The factors predisposing to hemolysis were not found, except the significantly diminished
Kalliopi Makarona et al.
Blood, 124(1), 134-141 (2014-05-09)
HDAC inhibitors (HDACi) increase transcription of some genes through histone hyperacetylation. To test the hypothesis that HDACi-mediated enhanced transcription might be of therapeutic value for inherited enzyme deficiency disorders, we focused on the glycolytic and pentose phosphate pathways (GPPPs). We
Elina Molou et al.
Scandinavian journal of clinical and laboratory investigation, 74(3), 259-263 (2014-01-28)
Glucose-6-Phosphate Dehydrogenase (G6PD) gene is located at the X-chromosome at Xq28 and the disease is recessively inherited predominantly in males. More than 400 variants have been proposed based on clinical and enzymatic studies. The aim of the current study was
Alphaxard Manjurano et al.
PLoS genetics, 11(2), e1004960-e1004960 (2015-02-12)
X-linked Glucose-6-phosphate dehydrogenase (G6PD) A- deficiency is prevalent in sub-Saharan Africa populations, and has been associated with protection from severe malaria. Whether females and/or males are protected by G6PD deficiency is uncertain, due in part to G6PD and malaria phenotypic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service