Skip to Content
Merck
All Photos(2)

Key Documents

A9521

Sigma-Aldrich

Anti-Glucose-6-Phosphate Dehydrogenase (G-6-PDH) antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

yeast

technique(s)

immunoelectrophoresis: suitable
indirect ELISA: 1:15,000-1:30,000

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

G-6-PDH catalyzes the NADP+(or NAD+)-dependent oxidation of D-glucose-6-phosphate to 6-phospho-D-glucono-1,5-lactone. This reaction is the rate limiting step of the pentose phosphate pathway.
Yeast G-6-PDH is often used as a loading control in several immunoassays.
Glucose-6-phosphate dehydrogenase (G6PD) is a cytosolic enzyme. It synthesizes NADPH (nicotinamide adenine dinucleotide phosphate). The G6PD gene is located on human chromosome X. It spans 18.5 kb and has 13 exons.
The product is specific for G-6-PDH as determined by immunoelectrophoresis. No reaction is observed with other proteins in a crude Baker′s yeast extract.

Immunogen

G-6-PDH from Baker′s yeast S. cerevisiae.

Application

Anti-Glucose-6-Phosphate Dehydrogenase (G-6-PDH) antibody has been used to detect the immunoreactive mass of glyceraldehyde 3-phosphate dehydrogenase (GAPDH). It has also been used in western blotting.
Rabbit Anti-Glucose-6-Phosphate Dehydrogenase (G-6-PDH) antibody has been used for western blot assays. The antibody can also be used for indirect ELISA (1:15,000-1:30,000) and immunoelectrophoresis.
Yeast cell extracts were analyzed by western blot and equal loading was determined by probing with rabbit anti-Glucose-6-Phosphate Dehydrogenase antibody.

Biochem/physiol Actions

NADPH (nicotinamide adenine dinucleotide phosphate), produced by glucose-6-phosphate dehydrogenase (G6PD) is an electron donor molecule, that helps in neutralizing harmful oxidizing agents. Deficiency in G6PD shows symptoms, that includes neonatal jaundice and acute episodic hemolysis. This enzyme guards the red blood cells against oxidative challenges.

Physical form

Lyophilized from 0.01 M phosphate buffered saline.

Reconstitution

Reconstitute with 2mL deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytochemical flow analysis of intracellular G6 PD and aggregate analysis of mosaic G6 PD expression
Kalnoky M, et al.
European Journal of Haematology, 100(3), 294-303 (2018)
Ming Li et al.
The Journal of cell biology, 211(3), 639-652 (2015-11-04)
Cells must regulate the abundance and activity of numerous nutrient transporters in different organelle membranes to achieve nutrient homeostasis. As the recycling center and major storage organelle, lysosomes are essential for maintaining nutrient homeostasis. However, very little is known about
Dimitrios Zattas et al.
The Journal of biological chemistry, 291(23), 12105-12118 (2016-04-14)
Specific proteins are modified by ubiquitin at the endoplasmic reticulum (ER) and are degraded by the proteasome, a process referred to as ER-associated protein degradation. In Saccharomyces cerevisiae, two principal ER-associated protein degradation ubiquitin ligases (E3s) reside in the ER
A conserved C-terminal element in the yeast Doa10 and human MARCH6 ubiquitin ligases required for selective substrate degradation
Zattas D, et al.
The Journal of Biological Chemistry, 29(7), jbc-M116 (2016)
Molecular Hematopathology
Hematologic Pathology, 712-760 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service