Skip to Content
Merck
All Photos(1)

Documents

HMSK-1

Sigma-Aldrich

Visikol® HISTO-M Starter Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.47

shipped in

ambient

storage temp.

room temp

General description

With the Visikol HISTO tissue clearing approach you are now able to rapidly and easily image tissues in 3D without damaging the underlying cellular morphology. The Visikol HISTO approach has been designed to pair with immunolabeling and is reversible so that 3D histology can be followed up with traditional 2D histology. The Visikol HISTO tissue clearing approach for 3D tissue cultures is comprised of one reagent called Visikol HISTO-M.The Visikol HISTO-M Starter Kit combines 30 mL of Visikol HISTO-M with the four buffer solutions required to process 3D tissue cultures for immunolabeling.  All you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of 3D cell culture models

Application

  • Optical clearing agent in tissue imaging studies
  • Refractive index matching agent for microscopy

Visikol HISTO products are easy-to-use and ideal for rapid tissue clearing applications. Tissue clearing is completely reversible with no alteration to the tissue.

Features and Benefits

  • Render tissue samples transparent for imaging.
  • Achieve clearer, sharper images in antibody tissue staining applications.
  • The Visikol HISTO tissue clearing process does not affect prior immunolabeling.
  • Save time. The Visikol HISTO method is rapid relative to many popular current tissue clearing techniques.
  • Retain morphology and lipid content in your samples.
  • The HISTO tissue clearing method is easily reversible for follow up 2D histology/IHC or for 3D data validation against traditional histology.
  • Non-toxic.

Legal Information

HISTO-1 is a trademark of Visikol, Inc.
HISTO-M is a trademark of Visikol, Inc.
Visikol is a registered trademark of Visikol, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.

Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.

Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.

Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service