Skip to Content
Merck
All Photos(1)

Key Documents

H2779

Sigma-Aldrich

HistoChoice® Clearing Agent

greener alternative

alternative to toluene and xylene

Synonym(s):

dewaxing mixture for paraffin tissue sections

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

form

liquid

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

colorless

density

0.760-0.820 g/mL

application(s)

hematology
histology

greener alternative category

storage temp.

room temp

General description

HistoChoice® is a patented, complex mixture of agents that allow tissues to retain their structure, antigenic sites, and nucleic acid sites. It is a non-toxic, non-cross-linking fixative formulated to replace formaldehyde-based fixation. This formulation in non-citrus, contains no mercury, formaldehyde, or glutaraldehyde.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of formalin for tissue processing.

Application

HistoChoice® Clearing Agent has the following applications:
  • It is suitable as an alternative to toluene and xylene for dewaxing paraffin tissue sections. Prepared slides are suitable for antibody probing applications and in situ hybridizations.
  • It has been employed in a study to explain the recalcitrance of lignocellulosic plant cell walls to enzymatic degradation.
  • It has been used to develop a novel method to improve sensitivity for the localization of miRNA and their target transcripts in plants.
  • It has been used to study the correlation of cytoplasmic ADP-ribosylation levels with markers of patient outcome in human cancers.

Biochem/physiol Actions

Fixation with HistoChoice® works well in immunohistology applications where its non-cross-linking traits are conducive to epitope retrieval and identification by primary antibodies to extracellular matrix components.

Features and Benefits

  • Non-toxic, non-carcinogenic, and odorless.
  • Non-flammable.
  • Preserves native antigenic and nucleic acid sites.
  • Compatible with many organic mounting media.
  • Dissolves wax faster and evaporates slower than xylene.
  • Can be used in automated processors.
  • Tissues retain their original structure.

Legal Information

HistoChoice is a registered trademark of Amresco, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

142.0 °F

Flash Point(C)

61.1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytoplasmic ADP-ribosylation levels correlate with markers of patient outcome in distinct human cancers
Aimi, et al.
Modern Pathology (2021)
Marie-Françoise Devaux et al.
Frontiers in plant science, 9, 200-200 (2018-03-09)
Tracking enzyme localization and following the local biochemical modification of the substrate should help explain the recalcitrance of lignocellulosic plant cell walls to enzymatic degradation. Time-lapse studies using conventional imaging require enzyme labeling and following the biochemical modifications of biopolymers
Anita S Patel et al.
The Journal of investigative dermatology, 128(12), 2888-2893 (2008-06-13)
An increasing number of studies report that genus beta human papillomaviruses (HPVs) are associated with skin cancer, with suggestions of specificity for squamous cell carcinoma (SCC) of the skin. We have conducted a systematic examination of HPV DNA in tumors
Carmen J Marsit et al.
Carcinogenesis, 27(1), 112-116 (2005-07-01)
Tobacco smoking, certain occupational exposures, and exposure to inorganic arsenic in drinking water have been associated with the occurrence of bladder cancer. However, in these tumors the exposure-associated pattern of somatic alterations in genes in the causal pathway for disease
Alexander Kunz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 28(7), 1649-1658 (2008-02-15)
CD36, a class-B scavenger receptor involved in multiple functions, including inflammatory signaling, may also contribute to ischemic brain injury through yet unidentified mechanisms. We investigated whether CD36 participates in the molecular events underlying the inflammatory reaction that accompanies cerebral ischemia

Related Content

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service