Skip to Content
Merck
All Photos(1)

Key Documents

H7291

Sigma-Aldrich

Interleukin-4 human

IL-4, potency:, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

≤0.6 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 14-19 kDa (glycosylated)

packaging

pkg of 1 mg
pkg of 10 μg
pkg of 100 μg

manufacturer/tradename

HumanZyme

technique(s)

cell culture | mammalian: suitable

impurities

<1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL4(3565)

Looking for similar products? Visit Product Comparison Guide

General description

Interleukin-4 (IL-4) is a pleiotropic cytokine and a complex glycoprotein mapped to human chromosome 5q31.1. It is produced by mast cells, basophils and a subset of activated T cells.

Biochem/physiol Actions

Interleukin-4 (IL-4) interacts with interleukin 13 and is an interleukin-4 receptor ligand. It regulates IgE and IgG1 production by B cells. IL-4 favors glial and neuronal differentiation and regulates differentiation of naive cluster of differentiation CD4+ T cells to the T helper type 2 (TH2) cells. Polymorphism in the IL-4 gene is implicated in immune and inflammatory disorders. In cerebral infarction, IL-4 levels are elevated.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Preparation Note

HumanKine Interleukin-4 (IL-4), expressed in human HEK 293 cells, is a glycosylated monomer with an apparent molecular mass of 14-19 kDa due to glycosylation, which is absent when this cytokine is expressed in E. coli. Production in human HEK 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The activity is determined by the dose-dependent simulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-4: role in disease and regulation of production.
P Choi et al.
Clinical and experimental immunology, 113(3), 317-319 (1998-09-16)
Interleukin 4 Receptor
Muscle and Exercise Physiology, 13(4) (1998)
Muscle as an endocrine organ
Muscle and Exercise Physiology, 13(4) (2019)
Neurogenesis, Inflammation, and Mental Health
Inflammation and Immunity in Depression, 13(4) (2018)
Robert Y L Zee et al.
Human molecular genetics, 13(4), 389-396 (2003-12-19)
Candidate gene polymorphisms related to inflammation, thrombosis and lipid metabolism have been implicated in the development of ischemic stroke. Using DNA samples collected at baseline in a prospective cohort of 14 916 initially healthy American men, we genotyped 92 polymorphisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service