H6389
Methyl hexacosanoate
≥99% (capillary GC)
Synonym(s):
Hexacosanoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)24COOCH3
CAS Number:
Molecular Weight:
410.72
Beilstein:
1800770
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥99% (capillary GC)
form
powder
functional group
ester
lipid type
saturated FAs
shipped in
ambient
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC
InChI
1S/C27H54O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27(28)29-2/h3-26H2,1-2H3
InChI key
VHUJBYYFFWDLNM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Determination of very-long-chain fatty acids in plasma by a simplified gas chromatographic-mass spectrometric procedure.
Caruso U, Fowler B, Erceg M, Romano C.
Journal of Chromatography A, 562, 147-152 (1991)
Phytanic acid, pristanic acid, and very-long-chain fatty acid methyl esters measured simultaneously by capillary gas chromatography.
H M Harris et al.
Clinical chemistry, 35(4), 703-704 (1989-04-01)
María J Rodríguez-López et al.
Insects, 11(9) (2020-09-13)
A combination of biological control and host plant resistance would be desirable for optimally controlling the greenhouse whitefly, Trialeurodes vaporariorum in tomato crops. Whitefly settlement preference, oviposition, and survivorship were evaluated on ABL 10-4 and 'Moneymaker', two nearly-isogenic tomato lines
M J Paik et al.
Journal of chromatography. B, Biomedical sciences and applications, 760(1), 149-157 (2001-08-28)
Pattern recognition analysis on the levels of the very-long-chain fatty acids (VLCFAs) in plasma is described for the visual discrimination of X-linked adrenoleukodystrophy (X-ALD) patients from normal healthy group. Plasma VLCFA compositions of 58 normal subjects and 16 X-ALD patients
Karlheinz Grillitsch et al.
Biochimica et biophysica acta, 1838(7), 1889-1897 (2014-04-01)
Despite similarities of cellular membranes in all eukaryotes, every compartment displays characteristic and often unique features which are important for the functions of the specific organelles. In the present study, we biochemically characterized the plasma membrane of the methylotrophic yeast
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service