Merck
  • An Acylsucrose-Producing Tomato Line Derived from the Wild Species Solanum pimpinellifolium Decreases Fitness of the Whitefly Trialeurodes vaporariorum.
An Acylsucrose-Producing Tomato Line Derived from the Wild Species Solanum pimpinellifolium Decreases Fitness of the Whitefly Trialeurodes vaporariorum.
Insects (2020-09-13)
María J Rodríguez-López, Enrique Moriones, Rafael Fernández-Muñoz
ABSTRACT
A combination of biological control and host plant resistance would be desirable for optimally controlling the greenhouse whitefly, Trialeurodes vaporariorum in tomato crops. Whitefly settlement preference, oviposition, and survivorship were evaluated on ABL 10-4 and 'Moneymaker', two nearly-isogenic tomato lines with, and without, whitefly-resistance traits based on type IV leaf glandular trichomes derived from the tomato wild species Solanum pimpinellifolium, respectively. Significantly reduced preference of T. vaporariorum adult whiteflies for ABL 10-4 leaves was observed. Moreover, T. vaporariorum altered its abaxial-adaxial settling performance on leaves of ABL 10-4 plants. A significantly lower tendency to settle on abaxial leaf surface was observed in ABL 10-4 compared to Moneymaker plants. Furthermore, T. vaporariorum deposited fewer eggs and exhibited a significantly reduced egg to adult survivorship in ABL 10-4 than in Moneymaker plants. Therefore, reduced fitness and distorted performance were observed for T. vaporariorum on ABL 10-4 tomato plants supporting that type IV leaf glandular trichomes might protect them from this pest and, indirectly, from the viruses it transmits.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Supelco
Invertase from baker′s yeast (S. cerevisiae), 200-300 units/mg solid
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Methyl hexacosanoate, ≥99% (capillary GC)