Skip to Content
Merck
All Photos(5)

Key Documents

H3281

Sigma-Aldrich

Hematin porcine

Synonym(s):

Ferrihemate, Hematin porcine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H33N4O5Fe
CAS Number:
Molecular Weight:
633.49
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.61

biological source

Porcine

form

powder

storage temp.

2-8°C

SMILES string

Cc1c(CCC(O)=O)c2cc3c(CCC(O)=O)c(C)c4cc5nc(cc6c(C=C)c(C)c(cc1n2)n6[Fe](O)n34)c(C)c5C=C

InChI

1S/C34H34N4O4.Fe.H2O/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);;1H2/q;+3;/p-3/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-;;

InChI key

BMUDPLZKKRQECS-HXFTUNQESA-K

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Hematin porcine has been used as a:
  • polymerase chain reaction (PCR) inhibitor to evaluate its effect on the PowerQuant System used for detecting degraded DNA in samples
  • supplement in Haemophilus test medium to culture Haemophilus influenzae isolates for resistance studies
  • standard to determine the heme coefficient as a part of heme iron analysis using raw beef lung

Hematin was used in a study to test in vitro activities of the Rx-01 oxazolidinones against hospital and community pathogens. It was also used in a study to test in-vitro bacterial identification using fluorescence spectroscopy with an optical fiber system.

Biochem/physiol Actions

Hematin is a hydroxide of heme. It may be used to treat acute porphyric attacks. Hematin suppresses the hepatic aminolevulinic acid (ALA) function and replenishes cytochrome action by restoring the endogenous heme deficit. Early administration of hematin can prevent irreversible neuronal damage induced due to metabolic disorders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Peter Bergman et al.
PloS one, 6(8), e24394-e24394 (2011-09-14)
Statin treatment has been associated with a beneficial outcome on respiratory tract infections. In addition, previous in vitro and in vivo experiments have indicated favorable effects of statins in bacterial infections. The aim of the present study was to elucidate
Brianna Atto et al.
Pathogens (Basel, Switzerland), 9(4) (2020-03-29)
Nontypeable Haemophilus influenzae (NTHi) is a leading causative organism of opportunistic respiratory tract infections. However, there are currently no effective vaccination strategies, and existing treatments are compromised by antibiotic resistance. We previously characterized Haemophilus haemolyticus (Hh) strains capable of producing
Muriel Vayssier-Taussat et al.
PLoS pathogens, 6(6), e1000946-e1000946 (2010-06-16)
Bacterial pathogens typically infect only a limited range of hosts; however, the genetic mechanisms governing host-specificity are poorly understood. The alpha-proteobacterial genus Bartonella comprises 21 species that cause host-specific intraerythrocytic bacteremia as hallmark of infection in their respective mammalian reservoirs
Jing Liu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 22(50), 18001-18008 (2016-10-27)
Graphene composites with hemin and gold nanoparticles show a better performance for hydrogen peroxide decomposition compared to that of the three components alone or duplex/hybrid complexes. Our previous studies showed that the morphology of the Au nanoparticles may greatly influence
Disorders of Peripheral Nerves
Katirji B
Bradley and Daroff's Neurology in Clinical Practice, 1853-1929 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service