Skip to Content
Merck
All Photos(3)

Documents

258385

Sigma-Aldrich

Protoporphyrin IX disodium salt

≥90% (HPLC)

Synonym(s):

3,7,12,17-Tetramethyl-8,13-divinyl-2,18-porphinedipropionic acid disodium salt, Depocolin-S, Ooporphyrin disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32N4Na2O4
CAS Number:
Molecular Weight:
606.62
Beilstein:
5227034
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥90% (HPLC)

form

powder

λmax

406 nm

storage temp.

2-8°C

SMILES string

[Na+].[Na+].Cc1c(CCC([O-])=O)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C=C)c5C)c(C=C)c4C)c(C)c3CCC([O-])=O

InChI

1S/C34H34N4O4.2Na/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16,36-37H,1-2,9-12H2,3-6H3,(H,39,40)(H,41,42);;/q;2*+1/p-2/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-;;

InChI key

JZRNDUPVNXWZDY-HXFTUNQESA-L

Looking for similar products? Visit Product Comparison Guide

Application

Protoporphyrin disodium salt (PpIX) was used as a dye content for fluorescence spectroscopic characterization in a study. The cytotoxic effect of PpIX on sarcoma 180 solid tumors cells was reported in a study.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fluorescence spectroscopic characterization of salivary metabolites of oral cancer patients.
Yuvaraj M, et al.
Journal of Photochemistry and Photobiology. B, Biology, 130, 153-160 (2014)
Kiwamu Takahashi et al.
Anticancer research, 33(7), 2861-2866 (2013-06-20)
5-Aminolevulinic acid (ALA) is a precursor of heme. ALA is used as a photosensitive substance in photodynamic diagnosis (PDD) and photodynamic therapy (PDT) because heme metabolism is abnormal in tumor cells and a photosensitive metabolite of heme synthesis from ALA
Sonodynamic effects of protoporphyrin IX disodium salt on isolated sarcoma 180 cells
Liu Q, et al.
Ultrasonics, 45(1-4), 56-60 (2006)
Kana Unuma et al.
PloS one, 8(2), e56526-e56526 (2013-03-05)
Lipopolysaccharide (LPS)-induced myocardial dysfunction is caused, at least in part, by mitochondrial dysfunction. Mitochondrial dysfunction and the oxidative damage associated with it are scavenged through various cellular defense systems such as autophagy to prevent harmful effects. Our recent study has
Filipa L Sousa et al.
Genome biology and evolution, 5(1), 200-216 (2012-12-22)
An open question regarding the evolution of photosynthesis is how cyanobacteria came to possess the two reaction center (RC) types, Type I reaction center (RCI) and Type II reaction center (RCII). The two main competing theories in the foreground of

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service