Skip to Content
Merck
All Photos(5)

Key Documents

H2126

Sigma-Aldrich

Hyaluronidase from sheep testes

Type II, lyophilized powder, ≥300 units/mg solid

Synonym(s):

Hyaluronate 4-glycanohydrolase, Hyaluronoglucosaminidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

conjugate

conjugate (Glucosaminoglycan)

Quality Level

type

Type II

form

lyophilized powder

specific activity

≥300 units/mg solid

mol wt

55 kDa

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hyaluronidase is a hyaluronic acid-metabolizing enzyme. It is found in the venoms of snake, honeybee and hymenoptera. Hyaluronidase degrades hyaluronic acid (HA), which enhances membrane permeability, lowers viscosity and increases tissue permeability. It enhances the diffusion of local anesthetics, lowers the size of myocardial infarction, improves the effects of anticancer drugs and prevents perineal trauma during spontaneous delivery.

Application

Hyaluronidase from sheep testes has been used:
  • in flow cytometry
  • in the isolation of putative spermatogonial stem cells (SSCs)
  • to study the effects of magnetic stimulation in magnetic hydrogel properties
  • in immunohistochemistry to visualize collagen type II distribution
  • to expose group-III auditory neurons of Muller′s organ for patch-clamp recordings
  • for the degeneration of hydrogels

Biochem/physiol Actions

These enzymes randomly cleave β-N-acetylhexosamine-(1−4)-glycosidic bonds in hyaluronic acid, chondroitin, and chondroitin sulfates.

Unit Definition

One unit will degrade 0.75 μg of the polysaccharide hyaluronic acid per minute at pH 5.35 at 37 ºC (as measured by turbidimetric absorbance (λ: 600nm) when complexed with BSA after 45 minutes).

Physical form

Lyophilized powder containing lactose

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert B Struijk et al.
PloS one, 15(3), e0230253-e0230253 (2020-03-17)
Autologous transplantation of spermatogonial stem cells is a promising new avenue to restore fertility in infertile recipients. Expansion of the initial spermatogonial stem cell pool through cell culturing is a necessary step to obtain enough cells for effective repopulation of
Yield stress determines bioprintability of hydrogels based on gelatin-methacryloyl and gellan gum for cartilage bioprinting
Mouser VHM, et al.
Biofabrication, 8(3), 035003-035003 (2016)
3D bioprinting of methacrylated hyaluronic acid (MeHA) hydrogel with intrinsic osteogenicity
Poldervaart MT, et al.
PLoS ONE, 12(6), e0177628-e0177628 (2017)
The role of the mechanotransduction ion channel candidate Nanchung-Inactive in auditory transduction in an insect ear.
Warren B and Matheson T
The Journal of Neuroscience, 2310-2317 (2018)
Miscellaneous Drugs, Materials, Medical Devices and Techniques
Nan A
Side Effects of Drugs Annual, 37, 603-619 (2015)

Articles

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

See All

Protocols

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

To measure hyaluronidase activity, a turbidimetric determination assay is used at 600 nm. One unit of hyaluronidase activity will cause a change in absorbance of 0.330 per minute at pH 5.35 at 37 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service